Skip to Content
Merck
All Photos(1)

Key Documents

PZ0306

Sigma-Aldrich

PF-6808472

≥98% (HPLC)

Synonym(s):

4-{4-[4-(5-Cyclopropyl-1H-pyrazol-3-ylamino)-6-prop-2-ynylcarbamoyl-pyrimidin-2-yl]-piperazin-1-ylmethyl}-benzenesulfonyl fluoride, XO44

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00
25 MG
$547.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$135.00
25 MG
$547.00

About This Item

Empirical Formula (Hill Notation):
C25H27FN8O3S
CAS Number:
Molecular Weight:
538.60
UNSPSC Code:
12352200
NACRES:
NA.77

$135.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

F[S](=O)(=O)c1ccc(cc1)CN2CCN(CC2)c3nc(cc(n3)C(=O)NCC#C)Nc4n[nH]c(c4)C5CC5

InChI

1S/C25H27FN8O3S/c1-2-9-27-24(35)21-15-22(29-23-14-20(31-32-23)18-5-6-18)30-25(28-21)34-12-10-33(11-13-34)16-17-3-7-19(8-4-17)38(26,36)37/h1,3-4,7-8,14-15,18H,5-6,9-13,16H2,(H,27,35)(H2,28,29,30,31,32)

InChI key

ZBSPMOBILDLOCV-UHFFFAOYSA-N

Biochem/physiol Actions

PF-6808472 is a cell permeable covalent probe suitable for click chemistry designated to measure kinase selectivity in intact cells. PF-6808472 contains sulfonyl fluoride group, which react with conserved lysine residue within kinase ATP binding site.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Zhao et al.
Journal of the American Chemical Society, 139(2), 680-685 (2017-01-05)
Protein kinases comprise a large family of structurally related enzymes. A major goal in kinase-inhibitor development is to selectively engage the desired kinase while avoiding myriad off-target kinases. However, quantifying inhibitor interactions with multiple endogenous kinases in live cells remains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service