Skip to Content
Merck
All Photos(1)

Key Documents

PZ0247

Sigma-Aldrich

PF-04979064

≥98% (HPLC)

Synonym(s):

1,3-Dihydro-1-[1-[(2S)-2-hydroxy-1-oxopropyl]-4-piperidinyl]-3-methyl-8-(6-methyl-3-pyridinyl)-2H-imidazo[4,5-c][1,5]naphthyridin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00
25 MG
$493.00

$122.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$122.00
25 MG
$493.00

About This Item

Empirical Formula (Hill Notation):
C24H26N6O3
CAS Number:
Molecular Weight:
446.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$122.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CN(C(N1C2CCN(C([C@@H](O)C)=O)CC2)=O)C3=C1C4=NC(C5=CN=C(C)C=C5)=CC=C4N=C3

InChI

1S/C24H26N6O3/c1-14-4-5-16(12-25-14)18-6-7-19-21(27-18)22-20(13-26-19)28(3)24(33)30(22)17-8-10-29(11-9-17)23(32)15(2)31/h4-7,12-13,15,17,31H,8-11H2,1-3H3/t15-/m0/s1

InChI key

GACQNUHFDBEIQH-HNNXBMFYSA-N

Biochem/physiol Actions

PF-04979064 is a highly potent and selective PI3K/mTOR dual kinase inhibitor.
PF-04979064 is an orally available, potent and selective PI3K/mTOR dual kinase inhibitor that potently inhibits tumor growth in mouse xenografts.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service