Skip to Content
Merck
All Photos(1)

Key Documents

CDS023114

Sigma-Aldrich

Aliskiren

Synonym(s):

(2S,4S,5S,7S)-5-Amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-4-hydroxy-2-isopropyl-7-[4-methoxy-3-(3-methoxypropoxy)benzyl]-8-methylnonanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H53N3O6
CAS Number:
Molecular Weight:
551.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

COCCCOC1=C(C=CC(C[C@H](C(C)C)C[C@@H]([C@H](C[C@H](C(NCC(C)(C(N)=O)C)=O)C(C)C)O)N)=C1)OC

InChI

1S/C30H53N3O6/c1-19(2)22(14-21-10-11-26(38-8)27(15-21)39-13-9-12-37-7)16-24(31)25(34)17-23(20(3)4)28(35)33-18-30(5,6)29(32)36/h10-11,15,19-20,22-25,34H,9,12-14,16-18,31H2,1-8H3,(H2,32,36)(H,33,35)/t22-,23-,24-,25-/m0/s1

InChI key

UXOWGYHJODZGMF-QORCZRPOSA-N

Gene Information

human ... REN(5972)

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Henrik H Hansen et al.
Drug discovery today, 22(11), 1707-1718 (2017-07-09)
Nonalcoholic fatty liver disease (NAFLD) has become the most common cause of chronic liver disease in the Western world. NAFLD is a complex spectrum of liver diseases ranging from benign hepatic steatosis to its more aggressive necroinflammatory manifestation, nonalcoholic steatohepatitis
Craig Balmforth et al.
JACC. Heart failure, 7(6), 457-465 (2019-05-13)
The purpose of this study was to compare outcomes (and the effect of sacubitril/valsartan) according to etiology in the PARADIGM-HF (Prospective comparison of angiotensin-receptor-neprilysin inhibitor [ARNI] with angiotensin-converting-enzyme inhibitor [ACEI] to Determine Impact on Global Mortality and morbidity in Heart
Elena Martínez-Klimova et al.
Biomolecules, 9(4) (2019-04-11)
Renal fibrosis is the common pathway for most forms of progressive renal disease. The Unilateral Ureteral Obstruction (UUO) model is used to cause renal fibrosis, where the primary feature of UUO is tubular injury as a result of obstructed urine
Takashi Miyake et al.
Hypertension (Dallas, Tex. : 1979), 70(6), 1201-1209 (2017-10-25)
Angiotensin II is thought to participate in aneurysm formation, because of its ability to induce and perpetuate inflammation in the aortic wall. Because activation of renin is the first step of the renin-angiotensin system, renin inhibition could inhibit all components
Amal M Mahfoz et al.
Naunyn-Schmiedeberg's archives of pharmacology, 389(12), 1315-1324 (2016-09-11)
Since chronic kidney disease due to diabetic nephropathy (DN) is becoming an ever larger health burden worldwide, more effective therapies are desperately needed. In the present study, the anti-diabetic and renoprotective effects of aliskiren have been evaluated in streptozotocin (STZ)-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service