Skip to Content
Merck
All Photos(1)

Key Documents

PZ0188

Sigma-Aldrich

PF-3644022 hydrate

≥98% (HPLC)

Synonym(s):

(10R)-9,10,11,12-tetrahydro-10-methyl-3-(6-methyl-3-pyridinyl)-8H-[1,4]diazepino[5′,6′:4,5]thieno[3,2-f]quinolin-8-one hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$185.00
25 MG
$705.00

$185.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$185.00
25 MG
$705.00

About This Item

Empirical Formula (Hill Notation):
C21H18N4OS · xH2O
Molecular Weight:
374.46 (anhydrous basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$185.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

O.C[C@@H]1CNc2c(sc3ccc4nc(ccc4c23)-c5ccc(C)nc5)C(=O)N1

InChI

1S/C21H18N4OS.H2O/c1-11-3-4-13(10-22-11)15-6-5-14-16(25-15)7-8-17-18(14)19-20(27-17)21(26)24-12(2)9-23-19;/h3-8,10,12,23H,9H2,1-2H3,(H,24,26);1H2/t12-;/m1./s1

InChI key

WHVUIWUUCVXAGR-UTONKHPSSA-N

Application

PF-3644022 hydrate has been used for the inhibition of mitogen-activated protein kinase 2 (MK2) in human osteosarcoma cells (U2OS)[1] and in colorectal carcinoma cells (CT26).[2]

Biochem/physiol Actions

PF-3644022 hydrate is a MK2 inhibitor
PF-3644022 is a potent inhibitor of mitogen-activated protein kinase-activated protein kinase 2 (MK2; Ki = 3 nM). PF-3644022 inhibits TNFa and IL-6 production in LPS-stimulated human whole blood (IC50 = 1.6 and 10.3 μM, respectively) and blocks TNFα production and paw swelling in a streptococcal cell wall-induced arthritis in rats.

Features and Benefits

This compound is featured on the MAPKAPs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

PF3644022 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the PF3644022 probe summary on the Chemical Probes Portal website.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oncogenic RAS signaling promotes tumor immunoresistance by stabilizing PD-L1 mRNA.
Coelho MA, et al.
Immunity, 47(6), 1083-1099 (2017)
Induction of oxidative metabolism by the p38alpha/MK2 pathway.
Trempolec N, et al.
Scientific Reports, 7(1), 11367-11367 (2017)
Philipp Trulley et al.
Cell reports, 27(10), 2859-2870 (2019-06-06)
Alternative translation is an important mechanism of post-transcriptional gene regulation leading to the expression of different protein isoforms originating from the same mRNA. Here, we describe an abundant long isoform of the stress/p38MAPK-activated protein kinase MK2. This isoform is constitutively
Natalia Ronkina et al.
Journal of immunology (Baltimore, Md. : 1950), 203(8), 2291-2300 (2019-09-19)
Tristetraprolin (TTP) is an RNA-binding protein and an essential factor of posttranscriptional repression of cytokine biosynthesis in macrophages. Its activity is temporally inhibited by LPS-induced p38MAPK/MAPKAPK2/3-mediated phosphorylation, leading to a rapid increase in cytokine expression. We compared TTP expression and
Nicholas Van Sciver et al.
PLoS pathogens, 17(11), e1010045-e1010045 (2021-11-09)
Epstein-Barr virus (EBV) is a human herpesvirus that causes infectious mononucleosis and contributes to both B-cell and epithelial-cell malignancies. EBV-infected epithelial cell tumors, including nasopharyngeal carcinoma (NPC), are largely composed of latently infected cells, but the mechanism(s) maintaining viral latency

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service