Merck
  • Alternative Translation Initiation Generates a Functionally Distinct Isoform of the Stress-Activated Protein Kinase MK2.
Alternative Translation Initiation Generates a Functionally Distinct Isoform of the Stress-Activated Protein Kinase MK2.
Cell reports (2019-06-06)
Philipp Trulley, Goda Snieckute, Dorte Bekker-Jensen, Manoj B Menon, Robert Freund, Alexey Kotlyarov, Jesper V Olsen, Manuel D Diaz-Muñoz, Martin Turner, Simon Bekker-Jensen, Matthias Gaestel, Christopher Tiedje
ABSTRACT
Alternative translation is an important mechanism of post-transcriptional gene regulation leading to the expression of different protein isoforms originating from the same mRNA. Here, we describe an abundant long isoform of the stress/p38MAPK-activated protein kinase MK2. This isoform is constitutively translated from an alternative CUG translation initiation start site located in the 5' UTR of its mRNA. The RNA helicase eIF4A1 is needed to ensure translation of the long and the known short isoforms of MK2, of which the molecular properties were determined. Only the short isoform phosphorylated Hsp27 in vivo, supported migration and stress-induced immediate early gene (IEG) expression. Interaction profiling revealed short-isoform-specific binding partners that were associated with migration. In contrast, the long isoform contains at least one additional phosphorylatable serine in its unique N terminus. In sum, our data reveal a longer isoform of MK2 with distinct physiological properties.
Sigma-Aldrich
PF-3644022 hydrate, ≥98% (HPLC)
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse