Skip to Content
Merck
All Photos(1)

Key Documents

PZ0136

Sigma-Aldrich

CP-100263 dihydrochloride hydrate

≥97% (HPLC)

Synonym(s):

(-)-CP 99994 dihydrochloride hydrate, (2R,3R)-N-[(2-Methoxyphenyl)methyl]-2-phenyl-3-piperidinamine dihydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$124.00
25 MG
$657.00

$124.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$124.00
25 MG
$657.00

About This Item

Empirical Formula (Hill Notation):
C19H24N2O·2HCl
CAS Number:
Molecular Weight:
369.33
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$124.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

solid

color

white to off-white

solubility

H2O: ≥5 mg/mL

storage temp.

room temp

SMILES string

O.Cl.Cl.COc1ccccc1CN[C@@H]2CCCN[C@@H]2c3ccccc3

InChI

1S/C19H24N2O.2ClH.H2O/c1-22-18-12-6-5-10-16(18)14-21-17-11-7-13-20-19(17)15-8-3-2-4-9-15;;;/h2-6,8-10,12,17,19-21H,7,11,13-14H2,1H3;2*1H;1H2/t17-,19-;;;/m1.../s1

InChI key

KBQMYQWHOPCSMG-ONBPXQNSSA-N

Biochem/physiol Actions

CP-100263 is an NK-1 neurokinin receptor antagonist.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service