The only Pur-a-lyzer™ products that are shipped in a wet condition are those with the 1,000, 25,000 and 50,000 MWCO. All the other MWCO Pur-a-lyzer™ products are shipped dry. The composition of this storage buffer is proprietary information. The volume of buffer is appropriate to the size of the Pur-a-lyzer™.
PURX60100
Pur-A-Lyzer™ Maxi Dialysis Kit
Maxi 6000, capacity 0.1-3 mL, pkg of 100 Maxi dialysis tubes, MWCO 6-8 kDa
Synonym(s):
Dialyzers, High through put dialysis tubing, puralyzer
Select a Size
$1,720.00
Select a Size
About This Item
$1,720.00
Recommended Products
material
regenerated cellulose membrane
packaging
pkg of 100 Maxi dialysis tubes
capacity
0.1-3 mL
tube L × diam.
8 cm × 2.2 cm
pore size
6-8 kDa MWCO
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The Pur-A-Lyzer Dialysis Tube allows rapid, high performance extraction of macromolecules without contamination. It is ideal for purification of proteins, automated protein sequencing, peptide mapping and amino acid analysis.
Application
- Dialysis or buffer exchange of volumes between 10 mL and 20 mL
- Preparation of protein samples for MALDI-MS
- Sample concentration
- Large-scale protein dialysis such as antibodies and recombinant protein purification
- Peptide dialysis for molecules as small as 10 amino acids
- Virus-particle purification
- Removal of contaminating micro molecules
- Purification of tissue culture extracts
- Removal of salts, surfactants, solvents and detergents
- Complex formation studies (protein-protein, protein-DNA and protein-RNA)
- pH and buffer adjustment of sample solution, protein extract or cell extract
- High through- put dialysis, a single beaker can be used for several Pur-A-Lyzer tubes on one floating pad
For your convenience the Pur-A-Lyzer kits come in four sizes:
Mini: 10-250 μL
Midi: 50-800 μL
Maxi: 0.1-3 mL
Mega: 3-20 mL
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
Are Pur-A-Lyzer™ Maxi Dialysis Kits packaged with solution to hydrate the dialysis membranes?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service