Skip to Content
Merck
All Photos(3)

Documents

PRS4243

Sigma-Aldrich

Anti-IFN-β antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Interferon, beta 1, fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFNB1(3456)

General description

Interferon-β (IFN-β) is a cytokine that belongs to type I interferons. The IFN-β gene is mapped on the human chromosome at 9p21.3.

Immunogen

a 17 amino acid peptide from near the center of human IFN-b.

Biochem/physiol Actions

Interferon-β (IFN-β) plays a major role in antibacterial and antiviral defense. It is also involved in apoptosis, cell growth, and autoimmune disorders. It binds to multimeric cell receptors and successful binding results in a cascade of intracellular events.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4243.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amber Kaplan et al.
Journal of immunology (Baltimore, Md. : 1950), 198(10), 4036-4045 (2017-04-16)
Type I IFNs are a cytokine family essential for antiviral defense. More recently, type I IFNs were shown to be important during bacterial infections. In this article, we show that, in addition to known cytokine functions, IFN-β is antimicrobial. Parts
Dongdong Xu et al.
Bioactive materials, 35, 82-98 (2024-01-29)
Repetitive implant-related infections (IRIs) are devastating complications in orthopedic surgery, threatening implant survival and even the life of the host. Biofilms conceal bacterial-associated antigens (BAAs) and result in a "cold tumor"-like immune silent microenvironment, allowing the persistence of IRIs. To
L Runkel et al.
Pharmaceutical research, 15(4), 641-649 (1998-05-20)
Two recombinant IFN-beta products have been approved for the treatment of multiple sclerosis, a glycosylated form with the predicted natural amino acid sequence (IFN-beta-1a) and a non-glycosylated form that has a Met-1 deletion and a Cys-17 to Ser mutation (IFN-beta-1b).
Stephen Cunningham et al.
Clinical pharmacology and therapeutics, 78(6), 635-646 (2005-12-13)
Interferon IFN-beta is indicated for the treatment of multiple sclerosis. A significant proportion of patients show a poor clinical response to therapy. Type I interferon exerts its effect at least partially through interaction of specific transcription factors with interferon-stimulated response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service