PLA0150
Rabbit anti-CBX5 Antibody, Affinity Purified
Powered by Bethyl Laboratories, Inc.
Synonym(s):
HEL25, HP1, HP1 alpha, HP1 alpha homolog, HP1 alpha homolog (Drosophila), HP1-ALPHA, HP1A, HP1Hs alpha, HP1Hs-alpha, antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog; Drosophila), epididymis luminal protein 25, heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
grade
Powered by Bethyl Laboratories, Inc.
species reactivity
human
technique(s)
immunohistochemistry: 1:2,000- 1:10,000
immunoprecipitation (IP): 2- 5 μg/mg
western blot: 1:1,000-1:10,000
accession no.
NP_036249.1
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
Gene Information
rabbit ... CBX5(23468)
Immunogen
Physical form
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service