Skip to Content
Merck

Skip To

PHYTOMLS

Phytochemical Metabolite Library Of Standards

Supplied by IROA Technologies

Synonym(s):

Mass Spectrometry Metabolite Library, PHYOMLS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$8,500.00

About This Item

NACRES:
NA.28
UNSPSC Code:
41116107

$8,500.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

packaging

pkg of 4 plates

manufacturer/tradename

IROA Technologies

plate size

96 wells

shipped in

dry ice

storage temp.

−20°C

General description

Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds to allow multiple mixture to be analyzed per injection.

Application

PHYTOMLS (Phytochemical Metabolite Library of Standards) is a collection of high-quality phytochemicals produced by many edible plants. These are high purity (>95%) compounds supplied in an economical, ready-to-use format. These plant standards are most commonly used to provide retention times and spectra for these key metabolic compounds, help optimize mass spectrometry analytical protocols, and qualify and quantify mass spectrometry sensitivity and limit of detection.

Diets containing fruits, vegetables, whole grains, legumes, nuts and plant-based beverages include many bioactive phytochemicals such as carotenoids, phenolic acids, flavonoids, stilbenes, hydroxycinnamic acids, phytosterols and lignans. Through various physiological mechanisms these compounds have protective or disease preventative properties and many studies have been undertaken to reveal the health benefits of phytochemicals.

Features and Benefits

365 unique small molecule organic acid metabolites covering key metabolic pathways, including the following classes of compounds:
  • Terpenes
  • Phytosterols
  • Flavonoids
  • Phenolic acids
  • Tannins
  • Stilbenes
  • Lignans
  • Carotenoids

To view a complete list of compounds in the library for recent lots, download the plate maps below:

The complete plate map with Name, parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID are provided with the software download.

Preparation Note

Compounds are pre-weighed and provided at 5 μg per well, enough for multiple injections. Arranged in 96-well polypropylene plates with alphanumerically assigned positions. Discovery software is provided to help construct libraries, useful for metabolomic compound ID.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
MLSDiscovery software tool is provided to support the extraction, manipulation, and storage of the data generated when using the PHYTOMLS and other authentic libraries of metabolite standards supplied by IROA Technologies. Software comes with user manual and training videos.

To download the software package, visit sigma.com/mlsdiscovery.

To view the MLSDiscovery Video visit: MLSDiscovery Tutorial Video.

Legal Information

MLSDiscovery is a trademark of IROA Technologies LLC
PHYTOMLS is a trademark of IROA Technologies LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
GUTMLSAAPMLSOAMLS
packaging

pkg of 4 plates

packaging

pkg of 2 plates

packaging

pkg of 2 plates

packaging

pkg of 1 plates

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

manufacturer/tradename

IROA Technologies

form

liquid

form

-

form

-

form

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

plate size

96 wells

plate size

96 wells

plate size

96 wells

plate size

96 wells

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice


signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1A - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 - STOT SE 2 - STOT SE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Lungs, Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters