GUTMLS
Microbiome Metabolite Library Of Standards
Supplied by IROA Technologies
Synonym(s):
GUTMLS, Mass Spectrometry Metabolite Library
Select a Size
Select a Size
About This Item
Recommended Products
Quality Level
packaging
pkg of 2 plates
manufacturer/tradename
IROA Technologies
plate size
96 wells
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Application
The gut microbiota is colonized by approximately 500 different species of bacteria performing a host of useful functions including the production of vitamins and hormones, preventing the growth of harmful species and training the immune system. Many gut microbiome studies focus on dietary, disease or dysregulated metabolic pathways.
Features and Benefits
- Organic acids and derivatives
- Allyl sulfur compounds
- Steroids and steroid derivatives
- Benzene and substituted derivatives
- Azoles
- Flavonoids and isoflavonoids
- Fatty Acyls
- Lipids and lipid-like molecules
- Indoles and derivatives
- Organonitrogen compounds
- Imidazopyrimidines
- Organooxygen compounds
- Pyridines and derivatives
- Nucleoside and nucleotide analogues
To view a complete list of compounds in the library for recent lots, download the plate maps below:
The complete plate map with Name, parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID are provided with the software download.
Preparation Note
Other Notes
To download the software package, click here -MLSDiscovery software.
To view the MLSDiscovery Video visit: MLSDiscovery Tutorial Video.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3
Target Organs
Eyes,Central nervous system, Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
We provide small molecule metabolite libraries and kits with a comprehensive range of primary metabolite compounds covering key metabolic pathways for your metabolomics research.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service