Skip to Content
Merck
All Photos(2)

Documents

PHG0005

SAFC

BIS-TRIS propane

Pharma Manufacturing

Synonym(s):

1,3-Bis[tris(hydroxymethyl)methylamino]propane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[CH2NHC(CH2OH)3]2
CAS Number:
Molecular Weight:
282.33
Beilstein:
1786109
EC Number:
MDL number:
UNSPSC Code:
12161705
NACRES:
NA.21

biological source

synthetic

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals; tested

useful pH range

6.3-9.5

pKa (25 °C)

(1) 6.8, (2) 9.0

mp

164-165 °C (lit.)

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

OCC(CO)(CO)NCCCNC(CO)(CO)CO

InChI

1S/C11H26N2O6/c14-4-10(5-15,6-16)12-2-1-3-13-11(7-17,8-18)9-19/h12-19H,1-9H2

InChI key

HHKZCCWKTZRCCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

Bis Tris Propane is a unique biological buffer also referred to as a “Universal” buffer, which means the buffer capacity covers a wide range. Bis Tris Propane has two pKas of 6.3 and 9.5, which expands the buffering range from 5.0-10.5-making this a versatile buffer for biological systems. Bis Tris Propane is considered to be non-toxic to culture cell lines and provides high-solution clarity.

A review of DNA polymerase techniques lists bis tris propane as a suitable buffer for PCR methods as a universal buffer pH gradient is employed.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Francesca Spyrakis et al.
ChemMedChem, 9(7), 1501-1511 (2014-03-13)
Cystalysin from Treponema denticola is a pyridoxal 5'-phosphate dependent lyase that catalyzes the formation of pyruvate, ammonia, and sulfide from cysteine. It is a virulence factor in adult periodontitis because its reaction contributes to hemolysis, which sustains the pathogen. Therefore
Natalie Jayne Werling et al.
Human gene therapy methods, 26(3), 82-92 (2015-05-09)
Adeno-associated viral (AAV) vectors show great promise for gene therapy because of their excellent safety profile; however, development of robust dose-determining assays for AAV has presented a significant challenge. With the ultimate goal of future harmonization and standardization of AAV
Roland D Kersten et al.
ACS chemical biology, 10(11), 2501-2511 (2015-09-18)
Terpenes are ubiquitous natural chemicals with diverse biological functions spanning all three domains of life. In specialized metabolism, the active sites of terpene synthases (TPSs) evolve in shape and reactivity to direct the biosynthesis of a myriad of chemotypes for
Milya Davlieva et al.
Nucleic acids research, 43(9), 4758-4773 (2015-04-22)
LiaR is a 'master regulator' of the cell envelope stress response in enterococci and many other Gram-positive organisms. Mutations to liaR can lead to antibiotic resistance to a variety of antibiotics including the cyclic lipopeptide daptomycin. LiaR is phosphorylated in
Frank Jansen et al.
Biotechnology and applied biochemistry, 61(6), 646-654 (2014-03-01)
The plant polymer lignin is the greatest source of aromatic chemical structures on earth. Hence, the chemically diverse lignin monomers are valuable raw materials for fine chemicals, materials synthesis, and food and flavor industries. However, extensive use of this natural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service