Skip to Content
Merck
All Photos(4)

Key Documents

B9410

Sigma-Aldrich

BIS-TRIS propane

BioXtra, ≥99.0% (titration)

Synonym(s):

1,3-Bis[tris(hydroxymethyl)methylamino]propane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$443.00
100 G
$1,120.00
250 G
$2,780.00

$443.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$443.00
100 G
$1,120.00
250 G
$2,780.00

About This Item

Linear Formula:
CH2[CH2NHC(CH2OH)3]2
CAS Number:
Molecular Weight:
282.33
Beilstein:
1786109
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$443.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

product line

BioXtra

Assay

≥99.0% (titration)

form

powder

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

pH

10.0-12.0 (1 M in H2O)

useful pH range

6.3-9.5

pKa (25 °C)

(1) 6.8, (2) 9.0

mp

164-165 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

absorption

≤0.05 at 280 in H2O at 1 M
≤0.1 at 260 in H2O at 1 M

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

OCC(CO)(CO)NCCCNC(CO)(CO)CO

InChI

1S/C11H26N2O6/c14-4-10(5-15,6-16)12-2-1-3-13-11(7-17,8-18)9-19/h12-19H,1-9H2

InChI key

HHKZCCWKTZRCCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • A Capillary Electrophoresis UV Detection-Based Method for Global Genomic DNA Methylation Assessment in Human Whole Blood.: BIS-TRIS propane is used as a buffering agent in the capillary electrophoresis method to assess global DNA methylation levels in human whole blood. The buffer′s role is crucial in maintaining the stability and reproducibility of the electrophoretic conditions (Zinellu et al., 2019).

  • Low potential manganese ions as efficient electron donors in native anoxygenic bacteria.: BIS-TRIS propane is used to buffer the experimental conditions in studying the role of low potential manganese ions as electron donors in bacteria. The findings enhance our understanding of electron transfer processes in microbial systems (Deshmukh et al., 2018).

  • Evaluation of Global Genomic DNA Methylation in Human Whole Blood by Capillary Electrophoresis UV Detection.: This research uses BIS-TRIS propane in a capillary electrophoresis method to evaluate DNA methylation levels. The buffer′s properties ensure accurate and reliable detection, which is crucial for epigenetic studies (Zinellu et al., 2017).

Other Notes

Easily compare specifications for BIS-TRIS propane products with the BIS-TRIS propane specification table.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hamed Tahmasbi et al.
Journal of pharmaceutical and biomedical analysis, 171, 1-7 (2019-04-07)
Secreted recombinant activated clotting factor VII activated (rFVIIa) in cell culture media missing gamma-carboxyglutamic acid (Gla) domain as a result of failure in gamma-carboxylation or cell lysis is called Gla-domainless impurity which has less negative charge compared to native rFVIIa.
R Coronado et al.
The Journal of general physiology, 79(4), 529-547 (1982-04-01)
A collection of organic cations has been used to probe the gross structural features of the ionic diffusion pathway in a K+-selective channel from sarcoplasmic reticulum (SR). Channels were incorporated into planar phospholipid bilayer membranes, and single-channel currents were measured
R Tóth-Boconádi et al.
Biophysical journal, 78(6), 3170-3177 (2000-05-29)
Buffers change the electric signals of light-excited bacteriorhodopsin molecules in purple membrane if their concentration and the pH of the low-salt solution are properly selected. "Positive" buffers produce a positive component, and "negative" buffers a negative component in addition to
Joseph S Brunzelle et al.
Archives of biochemistry and biophysics, 474(1), 157-166 (2008-04-01)
The three-dimensional structure of the catalytically efficient beta-xylosidase from Selenomonas ruminantium in complex with competitive inhibitor 1,3-bis[tris(hydroxymethyl)methylamino]propane (BTP) was determined by using X-ray crystallography (1.3A resolution). Most H bonds between inhibitor and protein occur within subsite -1, including one between
P Gómez-Tagle et al.
Inorganic chemistry, 40(15), 3786-3796 (2001-07-10)
Potentiometric titrations of the mixtures of lanthanide(III) perchlorates and bis-Tris propane (BTP) reveal formation of dinuclear hydroxo complexes M2(BTP)2(OH)n(6-n), where M = La(III), Pr(III), Nd(III), Eu(III), Gd(III), and Dy(III) and n = 2, 4, 5, or 6, in the pH

Related Content

Discover high-purity biological buffers for production scale. Ensure stability, pH control, and consistent supply with our expert manufacturing solutions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service