Skip to Content
Merck
All Photos(1)

Documents

PG240200

Supelco

Controlled pore glass–uncoated

nominal diameter 240 Å, 120-200 mesh

Synonym(s):

Glass, controlled-pore

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41115716
NACRES:
SB.52

form

powder

technique(s)

LPLC: suitable

matrix

Silica

matrix active group

polymer

particle size

120-200 mesh

pore size

240 Å nominal diameter

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service