Skip to Content
Merck
All Photos(1)

Key Documents

P9088

Sigma-Aldrich

Phosphatase, Alkaline from shrimp

≥1,000 DEA units/mL, buffered aqueous glycerol solution

Synonym(s):

Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

buffered aqueous glycerol solution

mol wt

58 kDa

concentration

≥1,000 DEA units/mL

foreign activity

RNAse, DNAse, none detected

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will hydrolyze 1 μmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37 °C.

Physical form

Solution in 50% glycerol containing 25 mM Tris-HCl, pH 7.6, 1 mM MgCl2 and 0.1 mM ZnCl2

Analysis Note

Protein determined by biuret

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petr G Vikhorev et al.
Scientific reports, 7(1), 14829-14829 (2017-11-03)
Dilated cardiomyopathy (DCM) is an important cause of heart failure. Single gene mutations in at least 50 genes have been proposed to account for 25-50% of DCM cases and up to 25% of inherited DCM has been attributed to truncating
Andrew E Messer et al.
Frontiers in physiology, 8, 348-348 (2017-06-24)
Hypertrophic cardiomyopathy (HCM) is the most common single gene inherited cardiomyopathy. In cats (Felix catus) HCM is even more prevalent and affects 16% of the outbred population and up to 26% in pedigree breeds such as Maine Coon and Ragdoll.
Conor Crean et al.
Nucleic acids research, 36(3), 742-755 (2007-12-18)
The carbonate radical anion is a biologically important one-electron oxidant that can directly abstract an electron from guanine, the most easily oxidizable DNA base. Oxidation of the 5'-d(CCTACGCTACC) sequence by photochemically generated CO3*- radicals in low steady-state concentrations relevant to
Isabel Moller et al.
Glycoconjugate journal, 25(1), 37-48 (2007-07-17)
Antibody-producing hybridoma cell lines were created following immunisation with a crude extract of cell wall polymers from the plant Arabidopsis thaliana. In order to rapidly screen the specificities of individual monoclonal antibodies (mAbs), their binding to microarrays containing 50 cell
Lars Hansen et al.
American journal of human genetics, 92(4), 575-583 (2013-04-09)
PGAP2 encodes a protein involved in remodeling the glycosylphosphatidylinositol (GPI) anchor in the Golgi apparatus. After synthesis in the endoplasmic reticulum (ER), GPI anchors are transferred to the proteins and are remodeled while transported through the Golgi to the cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service