Skip to Content
Merck
All Photos(1)

Key Documents

P8014

Sigma-Aldrich

4α-Phorbol 12,13-didecanoate

solid

Synonym(s):

4α-Phorbol 12-13-dicaprinate, 4αPDD

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$256.00

$256.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$256.00

About This Item

Empirical Formula (Hill Notation):
C40H64O8
CAS Number:
Molecular Weight:
672.93
Beilstein:
2514324
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

$256.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

form

solid

Quality Level

color

white

solubility

DMSO: soluble
ethanol: soluble
methanol: soluble

storage temp.

−20°C

SMILES string

CCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)C(C=C(CO)C[C@]3(O)C2C=C(C)C3=O)C4C(C)(C)[C@]14OC(=O)CCCCCCCCC

InChI

1S/C40H64O8/c1-7-9-11-13-15-17-19-21-32(42)47-36-28(4)39(46)30(24-29(26-41)25-38(45)31(39)23-27(3)35(38)44)34-37(5,6)40(34,36)48-33(43)22-20-18-16-14-12-10-8-2/h23-24,28,30-31,34,36,41,45-46H,7-22,25-26H2,1-6H3/t28-,30+,31-,34-,36-,38+,39-,40-/m1/s1

InChI key

DGOSGFYDFDYMCW-OEFRVDPMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4α-Phorbol 12,13-didecanoate has been used as a transient receptor potential vanilloid 4 (TRPV4) agonist:
  • to study its effects on Madin-Darby canine kidney (MDCK) cells[1]
  • to study its effects on Ca2+ influx in mice fallopian tubal cells[2]
  • to study its effects on hind paw swelling in mice[3]

Biochem/physiol Actions

4α-Phorbol 12,13-didecanoate is a transient receptor potential vanilloid 4 (TRPV4) agonist. It promotes extracellular calcium influx into the cells.[4]

Caution

Photosensitive

Reconstitution

Stock solution cannot be made in aqueous media; dissolve in a water-miscible organic solvent before dilution to working concentrations in aqueous media.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tumor-promoting activity of phorbol and four diesters of phorbol in mouse skin.
W M Baird et al.
Cancer research, 31(8), 1074-1079 (1971-08-01)
Y Fukushima et al.
The Biochemical journal, 320 ( Pt 1), 27-32 (1996-11-15)
The histamine H2 receptor is a member of the family of G-protein-coupled receptors and is linked to the activation of adenylate cyclase phospholipase C (PLC). In this study we examined the effects of protein kinase C (PKC) activation in Chinese
Yasuhito Motojima et al.
Neuroscience letters, 678, 76-82 (2018-05-08)
Pain management remains a major concern regarding the treatment of postoperative patients. Transient receptor potential (TRP) channels are considered to be new therapeutic targets for pain control. We investigated whether the genes Trpv1 and Trpv4 are involved in hind paw
M C LaPointe et al.
Molecular endocrinology (Baltimore, Md.), 7(10), 1284-1296 (1993-10-01)
During left ventricular hypertrophy, brain natriuretic peptide (BNP) and atrial natriuretic factor (ANF) mRNA levels increase, possibly due to stretch-induced activation of protein kinase C. Phorbol ester treatment of primary cultures of neonatal rat ventricular cardiocytes represents an in vitro
Jennifer K Lee et al.
Journal of biomechanics, 47(15), 3712-3718 (2014-12-03)
Osteoarthritis, a degenerative disease of the load-bearing joints, greatly reduces quality of life for millions of Americans and places a tremendous cost on the American healthcare system. Due to limitations of current treatments, tissue engineering of articular cartilage may provide

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service