Skip to Content
Merck
All Photos(4)

Key Documents

I9657

Sigma-Aldrich

Ionomycin from Streptomyces conglobatus

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$250.00
5 MG
$1,000.00

$250.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$250.00
5 MG
$1,000.00

About This Item

Empirical Formula (Hill Notation):
C41H72O9
CAS Number:
Molecular Weight:
709.00
Beilstein:
3642126
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

$250.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

biological source

Streptomyces conglobatus

Quality Level

Assay

≥98% (HPLC)

solubility

acetonitrile: 10 mg/mL

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

C[C@H](CCC(O)=O)C[C@H](C)C[C@H](C)C(=O)\C=C(/O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]2CC[C@](C)(O2)[C@@H](C)O

InChI

1S/C41H72O9/c1-25(21-29(5)34(43)24-35(44)30(6)22-27(3)20-26(2)14-15-38(46)47)12-11-13-28(4)39(48)31(7)36(45)23-33-16-18-41(10,49-33)37-17-19-40(9,50-37)32(8)42/h11,13,24-33,36-37,39,42-43,45,48H,12,14-23H2,1-10H3,(H,46,47)/b13-11+,34-24-/t25-,26-,27+,28-,29-,30+,31+,32-,33+,36+,37-,39-,40+,41+/m1/s1

InChI key

PGHMRUGBZOYCAA-ADZNBVRBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ionomycin from Streptomyces conglobatus has been used:
  • in magnesium-ionomycin treatment experiments
  • as a supplement in calcium-free Dulbecco′s modified Eagle medium (DMEM) for ionophore treatment
  • in the restimulation of peripheral blood mononuclear cells (PBMCs) to analyze cytokine production using flow cytometry (FCM)
  • as a calcium ionophore to treat oocytes to perform intracytoplasmic sperm injection (ICSI) combined with assisted oocyte activation (AOA)

Biochem/physiol Actions

Ionomycin is a polyether antibiotic and an ionophore produced by the bacterium Streptomyces conglobatus. This Ca2+ ionophore is more effective than A23187 as a mobile ion carrier for Ca2+. It is non-fluorescent. It induces apoptotic degeneration of embryonic cortical neurons. Ionomycin has been shown to induce central demyelination, inhibit adrenal bovine TREK-1 channels, and regulate cell division of mature human B cells. It is used to study the effects of calcium flux on endoplasmic reticulum (ER) stress, mitochondrial stress, and intrinsic apoptosis mechanisms. It also stimulates the intracellular production of cytokines, interferon, perforin, interleukin-2 (IL-2), and IL-4 usually in conjunction with PMA.
Ionomycin is highly specific for divalent cations. It may have antiproliferative effects on the human bladder cancer cell line both in vitro and in vivo. Ionomycin functions as an apoptosis inducer which was suggested to act by activation of a latent, calcium-responsive endonuclease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lauren M Sparks et al.
PloS one, 6(7), e21068-e21068 (2011-07-16)
Disturbances in lipid metabolism are strongly associated with insulin resistance and type 2 diabetes (T2D). We hypothesized that activation of cAMP/PKA and calcium signaling pathways in cultured human myotubes would provide further insight into regulation of lipid storage, lipolysis, lipid
Davina Bonte et al.
Fertility and sterility, 112(2), 266-274 (2019-05-28)
To investigate the extent to which assisted oocyte activation (AOA) improves clinical outcomes in patients diagnosed with oocyte activation deficiencies (OADs). Retrospective cohort study comparing AOA cycles and previous intracytoplasmic sperm injection (ICSI) cycles in couples experiencing low or total
Ye Tian et al.
BMC women's health, 20(1), 246-246 (2020-11-06)
Fertilization failure after intracytoplasmic sperm injection continues to affect couples and the etiology is not well-understood. We characterized a couple with 2-year history of primary unexplained infertility. Three different assisted reproduction attempts (IVF + rescue ICSI, ICSI and ICSI-AOA) showed repeated fertilization
Pierre Gantner et al.
Nature communications, 11(1), 4089-4089 (2020-08-17)
Clonal expansions occur in the persistent HIV reservoir as shown by the duplication of proviral integration sites. However, the source of the proliferation of HIV-infected cells remains unclear. Here, we analyze the TCR repertoire of single HIV-infected cells harboring translation-competent
Andrew J M Howden et al.
Nature methods, 10(4), 343-346 (2013-03-12)
Here we demonstrate quantitation of stimuli-induced proteome dynamics in primary cells by combining the power of bio-orthogonal noncanonical amino acid tagging (BONCAT) and stable-isotope labeling of amino acids in cell culture (SILAC). In conjunction with nanoscale liquid chromatography-tandem mass spectrometry

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service