Skip to Content
Merck
All Photos(1)

Documents

P5372

Sigma-Aldrich

Anti-Potassium Channel Kv7.1 (KCNQ1) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-KVLQT1, Anti-Voltage-gated potassium channel QKT subfamily member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, human

technique(s)

western blot: 1:200 using rat heart membranes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNQ1(3784)
mouse ... Kcnq1(16535)
rat ... Kcnq1(84020)

Specificity

Does not cross react with other QKT proteins.

Immunogen

synthetic peptide corresponding to amino acid residues 661-676 of human KCNQ1. This sequence has 14/16 residues identical in rat and mouse.

Target description

Potassium Channel Kv7.1 (KCNQ1) encodes a protein for a voltage-gated potassium channel required for the repolarization phase of the cardiac action potential. The gene product can form heteromultimers with two other potassium channel proteins,

Physical form

Lyophilized powder from phosphate buffered saline containing 1% bovine serum albumin and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service