Skip to Content
Merck
All Photos(1)

Key Documents

P4530

Sigma-Aldrich

Protamine-Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500

form

saline suspension

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

~50 μg/mL binding capacity (DNA)

storage temp.

2-8°C

Application

Protamine-agarose is used in protein chromatography, affinity chromatography and specialty resins. Protamine-agarose has been used to determine that Pichia pastoris (methylotrophic yeast) provides a convenient heterologous system for the production of recombinant subunits of human type 2A protein phosphatase (PP2A). Protamine-agarose has also been used to purify and characterize a novel protamine kinase in HL60 cells as well as to study myocardial infarction.

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V B Lokeshwar et al.
The Journal of biological chemistry, 264(32), 19318-19326 (1989-11-15)
Treatment of Swiss mouse 3T3 cells and human epidermoid carcinoma A431 cells with protamine at 37 degrees C increased the 125I-epidermal growth factor (EGF) binding activity at 4 degrees C. The effect of protamine on the increase of 125I-EGF binding
L Boscá et al.
The Biochemical journal, 290 ( Pt 3), 827-832 (1993-03-15)
The structural changes following the binding to protein kinase C (PKC) of activators that promote its translocation to lipid environments were studied by far-u.v. c.d. and intrinsic fluorescence measurements of the protein. In the absence of activators, PKC contained 40%
Bruce M Brown et al.
Biochemistry, 41(46), 13526-13538 (2002-11-13)
In steps of protein purification of bovine retinal protein phosphatase 2A (PP2A), phosducin dephosphorylation activity peaks coelute with a PP2A enzyme complex, shown by peptide sequence analysis to contain a B' subunit, B56 epsilon. Other PP2A complexes with a slightly
J L Rosales et al.
Journal of cellular physiology, 164(3), 509-521 (1995-09-01)
The role of ras on protein kinase C (PKC) signaling was examined in two keratinocyte cell lines. Increasing the level of extracellular calcium from 0.15 mM to 1.0 mM induces some features of differentiation in the spontaneously immortalized HaCaT line
G M Walton et al.
Analytical biochemistry, 161(2), 425-437 (1987-03-01)
An efficient high yield three-step purification procedure for protein kinase C consisting of ion exchange, hydrophobic interaction, and substrate affinity chromatographies is described. Protein which appears homogeneous on sodium dodecyl sulfate-polyacrylamide gel electrophoresis contains amino acid sequences, predicted from cDNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service