Skip to Content
Merck
All Photos(1)

Key Documents

P4066

Potassium ferricyanide

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J-E Kim et al.
Oncogene, 36(2), 219-230 (2016-05-31)
The downstream events and target genes of p53 in the process of senescence are not fully understood. Here, we report a novel function of the forkhead transcription factor Foxp3, which is a key player in mediating T-cell inhibitory functions, in
Thomas C Jhou et al.
Neuron, 61(5), 786-800 (2009-03-17)
Separate studies have implicated the lateral habenula (LHb) or amygdala-related regions in processing aversive stimuli, but their relationships to each other and to appetitive motivational systems are poorly understood. We show that neurons in the recently identified GABAergic rostromedial tegmental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service