Skip to Content
Merck
All Photos(1)

Key Documents

60299

Sigma-Aldrich

Potassium hexacyanoferrate(III)

BioUltra, ≥99.0% (RT)

Synonym(s):

Potassium ferricyanide, Red prussiate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$98.40

$98.40


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$98.40

About This Item

Linear Formula:
K3Fe(CN)6
CAS Number:
Molecular Weight:
329.24
EC Number:
MDL number:
UNSPSC Code:
12171602
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent grade
form:
crystals
solubility:
H2O: 1 M at 20 °C, clear, very strong brownish-yellow

$98.40


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

reagent grade

product line

BioUltra

Assay

≥99.0% (RT)

form

crystals

impurities

insoluble matter, passes filter test

pH

6-9 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, very strong brownish-yellow

anion traces

chloride (Cl-): ≤50 mg/kg
hexacyanoferrate(II) ([Fe(CN)6]4-): ≤200 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤10 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤10 mg/kg
Pb: ≤20 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

no transmittance (UV) in H2O at 1 M

SMILES string

[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium hexacyanoferrate(III) (red prussiate) may be used as an electron acceptor in detection systems such as flow injection chemiluminescence and laser-induced fluorescence.

Other Notes

Electron acceptor, employed in systems involving electron transport, e.g. NADH-cytochrome b5 reductase[1][2]; assay of complex I[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detergent-solubilized NADH-cytochrome b5 reductase.
K Mihara et al.
Methods in enzymology, 52, 102-108 (1978-01-01)
Preparation and properties of NADH: ubiquinone oxidoreductase (complexI), EC 1.6.5.3.
Y Hatefi
Methods in enzymology, 53, 11-14 (1978-01-01)
Incorporation of microsomal electron-transfer components into liposomes: considerations for diffusion-limited reactions.
P Strittmatter et al.
Methods in enzymology, 52, 206-211 (1978-01-01)
Soichiro Ijiri et al.
Journal of chromatography. A, 1217(18), 3161-3166 (2010-03-23)
Rhodamine 110 (Rho110) has been used in the highly sensitive analysis of monosaccharides, as it reacts with the reducing carbonyl group of the saccharides. The monosaccharide derivatives were investigated by capillary electrophoresis with laser-induced fluorescence detection. The derivatization was performed
Mark A Jordan et al.
Water research, 47(2), 841-849 (2012-12-04)
Representative and fast monitoring of wastewater influent and effluent biochemical oxygen demand (BOD) is an elusive goal for the wastewater industry and regulatory bodies alike. The present study describes a suitable assay, which incorporates activated sludge as the biocatalyst and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service