Skip to Content
Merck
All Photos(1)

Key Documents

P3766

Sigma-Aldrich

Phorbol 12-myristate 13-acetate 4-O-methyl ether

Synonym(s):

MPMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H58O8
CAS Number:
Molecular Weight:
630.85
Beilstein:
6554604
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

storage temp.

−20°C

Quality Level

SMILES string

CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)C(C=C(CO)C[C@@]3(OC)C2C=C(C)C3=O)C4C(C)(C)[C@]14OC(C)=O

InChI

1S/C37H58O8/c1-8-9-10-11-12-13-14-15-16-17-18-19-30(40)44-33-25(3)36(42)28(31-34(5,6)37(31,33)45-26(4)39)21-27(23-38)22-35(43-7)29(36)20-24(2)32(35)41/h20-21,25,28-29,31,33,38,42H,8-19,22-23H2,1-7H3/t25-,28+,29-,31-,33-,35-,36-,37-/m1/s1

InChI key

VCWNAJUHTLWQQT-XCHBPHIQSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Weak phorbol ester agonist; very weak inflammatory and tumor promoting agent.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soroush Sharbati et al.
Microorganisms, 7(3) (2019-03-02)
Pathogenic mycobacteria are able to persist intracellularly in macrophages, whereas non-pathogenic mycobacteria are effectively combated and eliminated after their phagocytosis. It is known that TGF-β plays an important role in this context. Infection with pathogenic mycobacteria such as Mycobacterium tuberculosis
Simon Ittig et al.
PLoS pathogens, 8(5), e1002667-e1002667 (2012-05-10)
Capnocytophaga canimorsus is a usual member of dog's mouths flora that causes rare but dramatic human infections after dog bites. We determined the structure of C. canimorsus lipid A. The main features are that it is penta-acylated and composed of
Rosella Abeti et al.
Brain : a journal of neurology, 134(Pt 6), 1658-1672 (2011-05-28)
Alzheimer's disease is characterized by β-amyloid accumulation in the central nervous system. As β-amyloid is neurotoxic in culture, we have explored the mechanisms of toxicity in the search for therapeutic targets for Alzheimer's disease and now identify a key role
Anke Sichelstiel et al.
PloS one, 9(2), e98440-e98440 (2014-06-12)
For patients with chronic lung diseases, such as chronic obstructive pulmonary disease (COPD), exacerbations are life-threatening events causing acute respiratory distress that can even lead to hospitalization and death. Although a great deal of effort has been put into research
H Ling et al.
British journal of pharmacology, 161(8), 1763-1777 (2010-08-20)
Shogaols are reported to possess anti-inflammatory and anticancer activities. However, the antimetastatic potential of shogaols remains unexplored. This study was performed to assess the effects of shogaols against breast cancer cell invasion and to investigate the underlying mechanisms. The anti-invasive

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service