Skip to Content
Merck
All Photos(1)

Key Documents

P1494

Sigma-Aldrich

Monoclonal Anti-PMCA4 antibody produced in mouse

~1 mg/mL, clone JA9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Plasma Membrane Calcium ATPase 4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

JA9, monoclonal

form

buffered aqueous solution

species reactivity

human, rat

concentration

~1 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: 1:15,000 using human or rat tissue
western blot: 1:1,000 using human or rat tissue

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP2B4(493)

Immunogen

purified human erythrocyte PMCA.

Physical form

Solution in phosphate buffered saline containing 1 mg/ml bovine serum albumin and 0.05 % sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karolina Varga et al.
Cell calcium, 55(2), 78-92 (2014-01-21)
The expression of the plasma membrane Ca2+ ATPase (PMCA) isoforms is altered in several types of cancer cells suggesting that they are involved in cancer progression. In this study we induced differentiation of MCF-7 breast cancer cells by histone deacetylase
Randa Naffa et al.
Cancers, 13(6) (2021-04-04)
We demonstrated that the plasma membrane Ca2+ ATPase PMCA4b inhibits migration and metastatic activity of BRAF mutant melanoma cells. Actin dynamics are essential for cells to move, invade and metastasize, therefore, we hypothesized that PMCA4b affected cell migration through remodeling
Randa Naffa et al.
Cells, 9(5) (2020-05-18)
Metastatic melanoma is the most aggressive type of skin cancer. Previously, we identified the plasma membrane Ca2+ pump isoform 4b (PMCA4b or ATP2B4) as a putative metastasis suppressor in BRAF mutant melanoma cells. Metastasis suppressors are often downregulated in cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service