Skip to Content
Merck
All Photos(3)

Documents

SAE0105

Sigma-Aldrich

MT1 (Melatonin Receptor 1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in Sf9 cells

description

N-Terminal contains a strep tag II and 10X Histidine tag followed by a TEV protease cleavage site.

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

44.8 kDa

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

Melatonin receptor 1 (MT1) is a class A GPCR that modulates neuronal firing, arterial vasoconstriction, cell proliferation in cancer cells, and reproductive and metabolic functions.

Sequence

MASAWSHPQFEKGGGSGGGSGGSAWSHPQFEKGAHHHHHHHHHHENLYFQGQGNGSALPNASQPVLRGDGARPSWLASALACVLIFTIVVDILGNLLVILSVYRNKKLRNAGNIFVVSLAVADLVVAIYPYPLVLMSIFNNGWNLGYLHCQVSGFLMGLSVIGSIFNITGIAINRYCYICHSLKYDKLYSSKNSLCYVLLIWLLTLAAVLPNLRAGTLQYDPRIYSCTFAQSVSSAYTIAVVVFHFLVPMIIVIFCYLRIWILVLQVRQRVKPDRKPKLKPQDFRNFVTMFVVFVLFAICWAPLNFIGLAVASDPASMVPRIPEWLFVASYYMAYFNSCLNAIIYGLLNQNFRKEYRRIIVSLCTARVFFVDSSNDVADRVKWKPSPLMTNNNVVKVDSV

Preparation Note

Formulated in 25mM Na2HPO4 pH 8.0, 150mM NaCl, 0.86%/0.18% Sarkosyl/CHS.

Storage and Stability

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot into single use aliquots. Store remaining undiluted protein in aliquots at -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Stauch et al.
Nature, 569(7756), E6-E6 (2019-05-03)
Change history: In this Letter, the rotation signs around 90°, 135° and 15° were missing and in the HTML, Extended Data Tables 2 and 3 were the wrong tables; these errors have been corrected online.
Shinobu Yasuo et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(9), 2885-2889 (2009-03-06)
Melatonin transmits photoperiodic signals that regulate reproduction. Two melatonin receptors (MT1 and MT2) have been cloned in mammals and additional melatonin binding sites suggested, but the receptor that mediates the effects of melatonin on the photoperiodic gonadal response has not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service