Skip to Content
Merck
All Photos(2)

Key Documents

P1093

Sigma-Aldrich

Monoclonal Anti-Paxillin antibody produced in mouse

clone PXC-10, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

PXC-10, monoclonal

mol wt

antigen 68-40 kDa

contains

15 mM sodium azide

species reactivity

bovine, rat, human, chicken, hamster

technique(s)

immunocytochemistry: suitable
microarray: suitable
western blot: 1:500 using a cultured human cell line extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PXN(5829)
rat ... Pxn(360820)

General description

Monoclonal Anti-Paxillin (mouse IgG1 isotype) is derived from the PXC-10 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Paxillin (68 kDa), is a cytoskeletal component. It is found in the ends of actin stress fibers of the focal adhesions, but not in adherens junctions of the cells. The paxillin molecule has a single binding site for vinculin, and at least two binding sites for focal adhesion kinase (FAK), that are separated by an intervening sequence of 100 amino acids.

Specificity

Recognizes an epitope located in the LIM1 domain of the paxillin molecule.

Immunogen

C-terminal part of recombinant chicken paxillin (amino acids 305-559)

Application

Monoclonal Anti-Paxillin antibody produced in mouse has been used in immunoblotting and immunocytochemistry.

Biochem/physiol Actions

Paxillin interacts with several proteins including members of the src family of tyrosine kinases, the transforming protein v-crk, the cytoskeletal protein vinculin, and the tyrosine kinase, focal adhesion kinase (FAK). This interaction has suggested a function for paxillin as a molecular adaptor, responsible for the recruitment of structural and signaling molecules to focal adhesions.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paxillin: a crossroad in pathological cell migration
Lopez CAM, et al.
Journal of Hematology & Oncology, 10(1), 50-50 (2017)
Paxillin: a new vinculin-binding protein present in focal adhesions.
Turner CE, et al.
The Journal of cell biology, 111(3), 1059-1068 (1990)
Emmanuella Tsirimonaki et al.
Virus research, 115(2), 158-168 (2005-09-20)
Bovine papillomaviruses (BPVs) are oncogenic viruses. In cattle, BPV-1/2 is associated with urinary bladder cancer and BPV-4 with upper GI tract cancer. BPV E5 is a small hydrophobic protein localised in the endoplasmic reticulum (ER) and Golgi apparatus (GA). E5
Gokhan Bahcecioglu et al.
International journal of biological macromolecules, 122, 1152-1162 (2018-09-16)
In this study, porcine fibrochondrocyte-seeded agarose, methacrylated gelatin (GelMA), methacrylated hyaluronic acid (MeHA) and GelMA-MeHA blend hydrogels, and 3D printed PCL scaffolds were tested under dynamic compression for potential meniscal regeneration in vitro. Cell-carrying hydrogels produced higher levels of extracellular
Armen H Mekhdjian et al.
Molecular biology of the cell, 28(11), 1467-1488 (2017-04-07)
Metastasis requires tumor cells to navigate through a stiff stroma and squeeze through confined microenvironments. Whether tumors exploit unique biophysical properties to metastasize remains unclear. Data show that invading mammary tumor cells, when cultured in a stiffened three-dimensional extracellular matrix

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service