Skip to Content
Merck
All Photos(1)

Key Documents

P0373

Sigma-Aldrich

PSB 1115 potassium salt hydrate

≥95% (HPLC)

Synonym(s):

1-Propyl-8-(4-sulfophenyl)xanthine potassium salt hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$200.00

$200.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$200.00

About This Item

Empirical Formula (Hill Notation):
C14H13KN4O5S · xH2O
CAS Number:
Molecular Weight:
388.44 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$200.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CCCN1C(=O)Nc2nc([nH]c2C1=O)-c3ccc(cc3)S(=O)(=O)O[K]

InChI

1S/C14H14N4O5S.K/c1-2-7-18-13(19)10-12(17-14(18)20)16-11(15-10)8-3-5-9(6-4-8)24(21,22)23;/h3-6H,2,7H2,1H3,(H,15,16)(H,17,20)(H,21,22,23);/q;+1/p-1

InChI key

CJOWSBOERSTJAR-UHFFFAOYSA-M

Biochem/physiol Actions

PSB 1115 is a highly selective, water-soluble, human A2B adenosine receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luo Yan et al.
Journal of medicinal chemistry, 47(4), 1031-1043 (2004-02-06)
Many currently known antagonists for P2 purinergic receptors are anionic molecules bearing one or several phenylsulfonate groups. Among the P1 (adenosine) receptor antagonists, the xanthine phenylsulfonates are a potent class of compounds. Due to their high acidity, phenylsulfonates are negatively
Osama M Abo-Salem et al.
The Journal of pharmacology and experimental therapeutics, 308(1), 358-366 (2003-10-18)
Caffeine, an adenosine A1, A2A, and A2B receptor antagonist, is frequently used as an adjuvant analgesic in combination with nonsteroidal anti-inflammatory drugs or opioids. In this study, we have examined the effects of novel specific adenosine receptor antagonists in an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service