Skip to Content
Merck
All Photos(1)

Key Documents

SML1983

Sigma-Aldrich

PSB-603

≥98% (HPLC)

Synonym(s):

8-(4-(4-(4-Chlorophenyl)piperazide-1-sulfonyl)phenyl)-1-propylxanthine, 8-[4-[[4-(4-Chlorophenyl)-1-piperazinyl]sulfonyl]phenyl]-3,9-dihydro-1-propyl-1H-purine-2,6-dione, PSB 603, PSB603

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$164.00
25 MG
$665.00

$164.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$164.00
25 MG
$665.00

About This Item

Empirical Formula (Hill Notation):
C24H25ClN6O4S
CAS Number:
Molecular Weight:
529.01
UNSPSC Code:
12352200
NACRES:
NA.77

$164.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to brownish

solubility

DMSO: 2 mg/mL to clear

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(N1CCC)NC2=C(N=C(C3=CC=C(S(=O)(N4CCN(C5=CC=C(Cl)C=C5)CC4)=O)C=C3)N2)C1=O

InChI

1S/C24H25ClN6O4S/c1-2-11-31-23(32)20-22(28-24(31)33)27-21(26-20)16-3-9-19(10-4-16)36(34,35)30-14-12-29(13-15-30)18-7-5-17(25)6-8-18/h3-10H,2,11-15H2,1H3,(H,26,27)(H,28,33)

InChI key

OVHCTHHFOHMNFV-UHFFFAOYSA-N

Biochem/physiol Actions

A high-affinity, selective antagonist against A2b adenosine receptor (A2b-AR, A2bR).
PSB-603 is an adenosine receptor A2b antagonist (IC50 = 1.13 nM against 10 μM A2bR agonist NECA-induced calcium response in Jurkat T cells in the presence of 200 nM A2aR antagonist MSX-2) that targets A2bR with high affinity (KD = 0.403 nM/human, 0.457 nM/rat, 4.09 nM/mouse A2bR; Ki against 0.3 nM [3H]PSB-603 = 0.553 nM/human, 0.355 nM/rat, 0.265 nM/mouse A2bR) and selectivity, exhibiting little affinity toward A1, A2a, or A3 adenosine receptors by competition binding assay (Ki >10 μM against 0.4 nM [3H]DPCPX for human/rat A1R, 1 nM [3H]MSX-2 for human/mouse/rat A2aR, 10 nM [3H]NECA for human/mouse/rat A3R) with the exception of mouse A1R (Ki = 42.2 nM against 0.4 nM [3H]DPCPX).
PSB-603 is known to modify cellular metabolism and enhance cellular sensitivity for chemotherapy. Thus, PSB-603 is recognized to possess anticancer action, which has been studied in colorectal cancer.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Mølck et al.
Cancer letters, 383(1), 135-143 (2016-10-30)
Adenosine is a multifaceted regulator of tumor progression. It modulates immune cell activity as well as acting directly on tumor cells. The A The A PSB-603 alters cellular metabolism in colorectal cancer cells and increases their sensitivity to chemotherapy. Although
Mohamad Wessam Alnouri et al.
Purinergic signalling, 11(3), 389-407 (2015-07-02)
Adenosine receptors (ARs) have emerged as new drug targets. The majority of data on affinity/potency and selectivity of AR ligands described in the literature has been obtained for the human species. However, preclinical studies are mostly performed in mouse or
Denis Chang et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(44), 27502-27508 (2020-10-23)
Cyclic dinucleotides (CDNs) are secondary messengers used by prokaryotic and eukaryotic cells. In mammalian cells, cytosolic CDNs bind STING (stimulator of IFN gene), resulting in the production of type I IFN. Extracellular CDNs can enter the cytosol through several pathways
The A2b adenosine receptor antagonist PSB-603 promotes oxidative phosphorylation and ROS production in colorectal cancer cells via adenosine receptor-independent mechanism.
M?lck C, et al.
Cancer Letters, 383(1), 135-143 (2016)
Thomas Borrmann et al.
Journal of medicinal chemistry, 52(13), 3994-4006 (2009-07-03)
A new series of 1-alkyl-8-(piperazine-1-sulfonyl)phenylxanthines was designed, synthesized, and characterized in radioligand binding and functional assays at A(2B) adenosine receptors. A(2B) antagonists with subnanomolar affinity and high selectivity were discovered. The most potent compounds were 1-ethyl-8-(4-(4-(4-trifluoromethylbenzyl)piperazine-1-sulfonyl)phenyl)xanthine (24, PSB-09120, K(i) (human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service