Yes, this product includes a pH probe as well. The electrode is 6.5 inches long with a diameter of 0.47 inches and has a 1-meter cable.
Recommended Products
material
epoxy shaft
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 83033961
ORP measuring range
-1999-+1999 mV
pH measuring range
0.00-14.00 pH unit
temperature measuring range
0-100 °C
battery power
4 AAA, 250 hr life
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
I would like to ask if this product contains a pH probe too? Or it is just a meter without any probe?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service