Key Documents
OH30378541
Ohaus® Dissolved Oxygen Meter
Model ST400D-B
About This Item
UNSPSC Code:
41115611
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30378541
dissolved oxygen measuring range
0.00-20.0 mg/L (0 - 50 °C)
battery power
4 AAA, 250 hr life
General description
Large backlit LCD screen provides easy to read results - even in low light environments. Ideal for secure field use with a meter stand, wrist strap and durable IP54 housing.
Application
Field and lab Dissolved Oxygen measurement
Features and Benefits
99 measurement memory, last calibration data recall, auto barometric pressure measurement, auto temperature compensation with manual salinity compensation
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)