Skip to Content
Merck
All Photos(1)

Key Documents

Z127264

Cochranes atom centers for Minit molecular models

carbon, tetrahedral, black

Synonym(s):

chemistry model sets, molecular model sets

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$29.90

$29.90

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
100 EA
$29.90

About This Item

UNSPSC Code:
60104506
NACRES:
NB.17

$29.90

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


manufacturer/tradename

Cochranes 0321

color

black

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service