Skip to Content
Merck
All Photos(4)

Key Documents

N6506

Sigma-Aldrich

Nicotinamide hypoxanthine dinucleotide sodium salt

≥92%

Synonym(s):

Deamino DPN, Deamino NAD, Deaminodiphosphopyridine nucleotide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$197.00
100 MG
$817.00
500 MG
$2,840.00

$197.00


Estimated to ship onMay 24, 2025



Select a Size

Change View
25 MG
$197.00
100 MG
$817.00
500 MG
$2,840.00

About This Item

Empirical Formula (Hill Notation):
C21H26N6O15P2
CAS Number:
Molecular Weight:
664.41
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

$197.00


Estimated to ship onMay 24, 2025


Assay

≥92%

form

powder

storage temp.

−20°C

SMILES string

[Na].NC(=O)C1=CC=C[N](=C1)C2OC(COP(O)(=O)OP(O)(=O)OCC3OC(C(O)C3O)n4cnc5C(=O)N=CNc45)C(O)C2O

InChI

1S/C21H27N6O15P2.Na.H/c22-17(32)9-2-1-3-26(4-9)20-15(30)13(28)10(40-20)5-38-43(34,35)42-44(36,37)39-6-11-14(29)16(31)21(41-11)27-8-25-12-18(27)23-7-24-19(12)33;;/h1-4,7-8,10-11,13-16,20-21,28-31H,5-6H2,(H2,22,32)(H,34,35)(H,36,37)(H,23,24,33);;

InChI key

XVDVDRZCJWZVDL-UHFFFAOYSA-N

Related Categories

Application

Nicotinamide hypoxanthine dinucleotide (deamino-NAD) may be used study the specificity and kinetics of NADH: ubiquinone oxidoreductase(s).

Other Notes

Analog of β-NAD

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiwon Kang et al.
Journal of biochemistry and molecular biology, 40(1), 53-57 (2007-01-25)
The enzymatic properties of NADH:quinone oxidoreductase were examined in Triton X-100 extracts of Bacillus cereus membranes by using the artificial electron acceptors ubiquinone-1 and menadione. Membranes were prepared from B. cereus KCTC 3674 grown aerobically on a complex medium and
Rosa Pello et al.
Human molecular genetics, 17(24), 4001-4011 (2008-09-23)
Leber's hereditary optic neuropathy (LHON), the most frequent mitochondrial disorder, is mostly due to three mitochondrial DNA (mtDNA) mutations in respiratory chain complex I subunit genes: 3460/ND1, 11778/ND4 and 14484/ND6. Despite considerable clinical evidences, a genetic modifying role of the
Jukka Pätsi et al.
The Biochemical journal, 409(1), 129-137 (2007-09-27)
LHON (Leber hereditary optic neuropathy) is a maternally inherited disease that leads to sudden loss of central vision at a young age. There are three common primary LHON mutations, occurring at positions 3460, 11778 and 14484 in the human mtDNA
Kenji Kawahara et al.
Journal of biochemistry, 145(2), 229-237 (2008-12-09)
In the intraerythrocytic stages of malaria parasites, mitochondria lack obvious cristae and are assumed to derive energy through glycolysis. For understanding of parasite energy metabolism in mammalian hosts, we isolated rodent malaria mitochondria from Plasmodium yoelii yoelii grown in mice.
Pilvi Maliniemi et al.
Mitochondrion, 9(6), 394-401 (2009-07-21)
Seven of the 45 subunits of mitochondrial NADH:ubiquinone oxidoreductase (complex I) are mitochondrially encoded and have been shown to harbor pathogenic mutations. We modeled the human disease-associated mutations A4136G/ND1-Y277C, T4160C/ND1-L285P and C4171A/ND1-L289M in a highly conserved region of the fourth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service