N6377
Norharmane hydrochloride
crystalline
Synonym(s):
β-Carboline, 9H-Pyrido(3,4-b)indole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H8N2 · HCl
CAS Number:
Molecular Weight:
204.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
form
crystalline
color
yellow to green
storage temp.
2-8°C
SMILES string
Cl.c1ccc2c(c1)[nH]c3cnccc23
InChI
1S/C11H8N2.ClH/c1-2-4-10-8(3-1)9-5-6-12-7-11(9)13-10;/h1-7,13H;1H
InChI key
NEECSQBLLQEFLK-UHFFFAOYSA-N
Biochem/physiol Actions
Inhibitor of indoleamine 2,3-dioxygenase (IDO).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Monica M Arnold et al.
Neuropharmacology, 85, 293-304 (2014-06-08)
Tobacco use is the leading cause of preventable death. Although the health risks are well known, cessation rates remain low. Whereas behavioral and neuroanatomical studies on tobacco addiction conventionally use nicotine, there is evidence that other constituents, such as monoamine
B P Connop et al.
Neuroscience letters, 190(1), 69-72 (1995-04-28)
7-Nitro indazole (7-NI) has been used as a selective inhibitor of neuronal nitric oxide synthase (NOS) in vivo. This agent has a short duration of action which may be due to its metabolism. The structure of 7-NI resembles that of
Caiping Tan et al.
Dalton transactions (Cambridge, England : 2003), 40(34), 8611-8621 (2011-08-02)
Three novel Ru(II) complexes of the general formula [Ru(N-N)(2)(Norharman)(2)](SO(3)CF(3))(2), where N-N = 2,2'-bipyridine (bpy, 1), 1,10-phenanthroline (phen, 2), 4,7-diphenyl-1,10-phenanthroline (DIP, 3) and Norharman (9H-pyrido[3,4-b]indole) is a naturally occurring β-carboline alkaloid, have been synthesized and characterized. The molecular structures of 1
Tomás Herraiz et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(8), 1773-1781 (2011-05-11)
Monoamine oxidase (MAO) enzymes located in human mitochondria oxidize neurotransmitters and bioactivate the neurotoxin 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP) by oxidation to directly-acting neurotoxic pyridinium cations (MPDP⁺/MPP⁺) that produce Parkinsonism. Antioxidants and MAO inhibitors are useful as neuroprotectants. Naturally-occurring substances, antioxidants and redox
Ute Jautz et al.
Journal of agricultural and food chemistry, 56(12), 4311-4319 (2008-05-29)
A recently developed HPTLC/UV-FLD method was compared to the routinely used HPLC/UV-FLD method for the quantification of heterocyclic aromatic amines (HAA) formed at trace levels during the heating process of meat. For formation of these process contaminants under normal cooking
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service