Skip to Content
Merck
All Photos(4)

Key Documents

N6125

Sigma-Aldrich

Naphthol AS-TR phosphate disodium salt

fluorogenic, ≥99% (HPLC), powder

Synonym(s):

NASTRp disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$82.80
1 G
$229.00
5 G
$783.00

$82.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
100 MG
$82.80
1 G
$229.00
5 G
$783.00

About This Item

Empirical Formula (Hill Notation):
C18H13ClNNa2O5P
CAS Number:
Molecular Weight:
435.71
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

$82.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Product Name

Naphthol AS-TR phosphate disodium salt, ≥99% (HPLC), Bulk package

Assay

≥99% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Na+].[Na+].Cc1cc(Cl)ccc1NC(=O)c2cc3ccccc3cc2OP([O-])([O-])=O

InChI

1S/C18H15ClNO5P.2Na/c1-11-8-14(19)6-7-16(11)20-18(21)15-9-12-4-2-3-5-13(12)10-17(15)25-26(22,23)24;;/h2-10H,1H3,(H,20,21)(H2,22,23,24);;/q;2*+1/p-2

InChI key

TYCHZTPSWMGRRI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Substrate for the histochemical demonstration of acid and alkaline phosphatase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adéla Kratochvílová et al.
Scientific reports, 10(1), 7489-7489 (2020-05-06)
Caspases are proteases traditionally associated with inflammation and cell death. Recently, they have also been shown to modulate cell proliferation and differentiation. The aim of the current research was to search for osteogenic molecules affected by caspase inhibition and to specify
Claire Chevalier et al.
STAR protocols, 2(2), 100452-100452 (2021-04-30)
Mesenchymal-derived osteoblasts play a key role in bone formation via synthesis and mineralization of the bone and bone remodeling. Osteoclasts are multinucleated cells of hematopoietic origin with a role in bone resorption. Here, we describe a protocol for generating primary
Eva Svandova et al.
Journal of anatomy, 235(2), 256-261 (2019-05-31)
FASL (CD178) is known for its role in triggering apoptosis, mostly in relation with immune cells but additional functions have been reported more recently, including those in bone development. Examination of postnatal FasL-deficient mice (gld) showed an increased bone deposition
Barbora Vesela et al.
Frontiers in cell and developmental biology, 8, 589136-589136 (2020-11-13)
Caspases are evolutionary conserved proteases traditionally known as participating in apoptosis and inflammation but recently discovered also in association with other processes such as proliferation or differentiation. This investigation focuses on caspase-12, ranked among inflammatory caspases but displaying other, not
Claire Chevalier et al.
Cell metabolism, 32(4), 575-590 (2020-09-12)
Osteoporosis is the most prevalent metabolic bone disease, characterized by low bone mass and microarchitectural deterioration. Here, we show that warmth exposure (34°C) protects against ovariectomy-induced bone loss by increasing trabecular bone volume, connectivity density, and thickness, leading to improved

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service