Skip to Content
Merck
Naphthol AS-TR phosphate disodium salt ≥99% (HPLC), Bulk package

Naphthol AS-TR phosphate disodium salt

Naphthol AS-TR phosphate disodium salt ≥99% (HPLC), Bulk package
Empirical Formula (Hill Notation):
C18H13ClNNa2O5P
CAS Number:
Molecular Weight:
435.71

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing