Skip to Content
Merck
All Photos(1)

Key Documents

N4787

Sigma-Aldrich

Nek2, Active human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Never In Mitosis Gene A-Related Kinase 2, Nima-Related Kinase 2

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$734.00

$734.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
5 μG
$734.00

About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

$734.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

≥85% (SDS-PAGE)

form

buffered aqueous glycerol solution

enzyme activity

≥10 units/mg protein

mol wt

protein apparent mol wt ~80 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK2(4751)

Application

Kinase activity is measured as the molar amount of phosphate incorporated into myelin basic protein, per minute per mg protein at 30 °C using a final concentration of 50 μM [32P] ATP.

Biochem/physiol Actions

Nek2 (nimA-related kinase 2) is a cell cycle-regulated protein kinase with maximal activity at the onset of mitosis that localizes to the centrosome.

Unit Definition

One unit will incorporate 1 nanomole of phosphate into myelin basic protein (MBP) per minute at pH 7.2 at 30 deg C using a final concentration of 50 μm (32p) ATP

Physical form

Solution in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, and 30% glycerol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service