Skip to Content
Merck
All Photos(2)

Key Documents

N2286

Sigma-Aldrich

Nonactin

from Streptomyces griseus, ≥98.0% (Total homologs, HPLC)

Synonym(s):

Ammonium ionophore I, Ammonium ionophore

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$243.00
100 MG
$1,440.00

$243.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$243.00
100 MG
$1,440.00

About This Item

Empirical Formula (Hill Notation):
C40H64O12
CAS Number:
Molecular Weight:
736.93
Beilstein:
76434
EC Number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76

$243.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

Streptomyces griseus

Quality Level

description

Natural macrotetrolide, may contain homologues (like monactin and dinactin)

Assay

≥98.0% (Total homologs, HPLC)

form

powder

solubility

chloroform: soluble 10 mg/mL

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

DNA synthesis | interferes
cell membrane | interferes

storage temp.

2-8°C

SMILES string

C[C@H]1C[C@@H]2CC[C@@H](O2)[C@@H](C)C(=O)O[C@H](C)C[C@H]3CC[C@H](O3)[C@H](C)C(=O)O[C@@H](C)C[C@@H]4CC[C@@H](O4)[C@@H](C)C(=O)O[C@H](C)C[C@H]5CC[C@H](O5)[C@H](C)C(=O)O1

InChI

1S/C40H64O12/c1-21-17-29-9-13-34(49-29)26(6)38(42)46-23(3)19-31-11-15-36(51-31)28(8)40(44)48-24(4)20-32-12-16-35(52-32)27(7)39(43)47-22(2)18-30-10-14-33(50-30)25(5)37(41)45-21/h21-36H,9-20H2,1-8H3/t21-,22+,23+,24-,25-,26+,27+,28-,29-,30+,31+,32-,33-,34+,35+,36-

InChI key

RMIXHJPMNBXMBU-QIIXEHPYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Nonactin is a macrotetrolide that forms complexes with alkali cations, such as potassium and sodium. Nonactin has been reported to inhibit the processing of cytoplasmic precursor proteins destined for the mitochondria. Nonactin is an uncoupler of oxidative phosphorylation [1].

Biochem/physiol Actions

Nonactin is used as a neutral lipid membrane soluble ammonium (and potassium) ionophore, and to inhibit surface expression of certain heat shock proteins (HSP60) [2][3].

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei-Ji Zhao et al.
Archives of pharmacal research, 28(11), 1228-1232 (2005-12-15)
Four compounds, including two novel macrolides, were isolated from an endophyte Streptomyces sp. Is9131 of Maytenus hookeri. Spectral data indicated that these compounds were dimeric dinactin (1), dimeric nonactin (2), cyclo-homononactic acid (3), and cyclo-nonactic acid (4). Bioassay results showed
Total synthesis of nonactin.
Ju Y.L.; Byeang H.K.
ChemInform, 52, 571-571 (1996)
Brian R Kusche et al.
Journal of the American Chemical Society, 131(47), 17155-17165 (2009-11-12)
Nonactin is a polyketide antibiotic produced by Streptomyces griseus ETH A7796 and is an ionophore that is selective for K(+) ions. It is a cyclic tetraester generated from two monomers of (+)-nonactic acid and two of (-)-nonactic acid, arranged (+)-(-)-(+)-(-)
V Gh Shlyonsky et al.
Biochimica et biophysica acta, 1758(11), 1723-1731 (2006-10-31)
We describe the phenomenon of mechanoelectrical transduction in macroscopic lipid bilayer membranes modified by two cation-selective ionophores, valinomycin and nonactin. We found that bulging these membranes, while maintaining the membrane tension constant, produced a marked supralinear increase in specific carrier-mediated
Brian R Kusche et al.
Bioorganic & medicinal chemistry letters, 19(4), 1233-1235 (2009-01-27)
Nonactin, produced by Streptomyces griseus ETH A7796, is a macrotetrolide assembled from nonactic acid. It is an effective inhibitor of drug efflux in multidrug resistant erythroleukemia K562 cells at sub-toxic concentrations and has been shown to possess both antibacterial and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service