Key Documents
MTCH6800100
MTC Bio Halo Lightweight Single Channel Pipettor
capacity 10-100 μL
Synonym(s):
pipette
About This Item
UNSPSC Code:
41121500
NACRES:
NB.15
Pricing and availability is not currently available.
feature
adjustable
manual
no. of channels 1
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio H6800-100
capacity
10-100 μL
General description
As one of the lightest pipettors ever introduced, the Halo Pipettor dramatically reduces the risk of repetitive stress injury and the general shoulder, arm, and hand fatigue associated with daily pipetting. Reduced actuation forces andan ergonomic design make Halo Pipettors feel balanced and natural in hand. Featuring a four-digit micrometer display for precise volume selection and a click-based adjustment wheel with DriftGuard protection, the Halo Pipettor is perfect for applications that demand exceptional accuracy. With a recessed ejector button, and cushioned, shock-absorbing stop, the ergonomics of tip ejection have been re-imagined to relieve stress on the thumb and joints.
Halo Single Channel Pipettor, lightweight, 0.5-10µL
Features and Benefits
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Legal Information
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)