Skip to Content
Merck
All Photos(1)

Key Documents

MAK386

Sigma-Aldrich

Pyrophosphate (PPi) Assay Kit (Fluorometric/Colorimetric)

sufficient for 50 colorimetric or 100 fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

detection method

colorimetric
fluorometric

relevant disease(s)

genitourinary disorders; rheumatological diseases

storage temp.

−20°C

General description

Inorganic pyrophosphate (diphosphate, pyrophosphoric acid or PPi) is a small molecule formed during, or required as a substrate, for a number of biochemical reactions. Enzymatic reactions that generate PPi include ATP hydrolysis, DNA and RNA polymerization, cyclic AMP formation, and enzymatic activation of fatty acids to form their CoA esters. Conversely, enzymes that utilize PPi may include cyclases, hydrolases and ligases. Measurement of inorganic pyrophosphate (PPi) in plasma, serum and other biologic fluids is important in studies of bone metabolism, renal stone disease, and certain types of arthritis.

Features and Benefits

The Pyrophosphate (PPi) Assay Kit is:
  • Compatible with high-throughput handling systems.
  • Specific for Pyrophosphate: other molecules containing phosphate groups do not interfere with the assay.
  • Detects as little as 2 μM of Pyrophosphate in a variety of biological samples.

Suitability

The kit is suitable for the determination of PPi concentration, monitoring pyrophosphate release by a variety of enzymes and for the screening inhibition or activities of enzymes that consume pyrophosphate. Suitable samples include plasma, serum and other biological fluids, cell culture extracts and tissue lysates.

Principle

The Pyrophosphate (PPi) Assay Kit provides a fast, convenient and ultrasensitive method for determination of free inorganic pyrophosphate levels in biological material. PPi produced during biotic processes is detected through a series of reactions which utilize a proprietary enzyme mix and probe, generating a stable product that can be quantified by either colorimetric or fluorometric readout. Generated fluorescence (λEx = 535 nm/λEm = 587 nm) or color (A570 nm) intensities are directly proportional to the concentrations of pyrophosphate, enabling precise measurements. Monomeric inorganic phosphate (Pi) does not interfere with the assay.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service