Skip to Content
Merck
All Photos(2)

Key Documents

MAK169

Sigma-Aldrich

High Sensitivity Pyrophosphate Assay Kit

sufficient for 200 fluorometric tests

Synonym(s):

Pyrophosphate (PPi) Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$789.00

$789.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 KIT
$789.00

About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

$789.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

usage

sufficient for 200 fluorometric tests

application(s)

cosmetics
food and beverages

detection method

fluorometric

relevant disease(s)

genitourinary disorders; rheumatological diseases

storage temp.

−20°C

General description

Pyrophosphate (PPi) is produced by a number of biochemical reactions such as ATP hydrolysis, DNA and RNA polymerizations, cyclic AMP formation, and the formation of fatty acid-coenzyme A esters.

Suitability

The Pyrophosphate Assay Kit provides a simple and direct flourescent microplate procedure for measuring pyrophosphate in a variety of samples.

Principle

The pyrophosphate concentration of a sample is determined by the use of a unique fluorogenic pyrophosphate sensor in which the presence of pyrophosphate results in the production of a fluorescent product (λex =370/λem = 470 nm) proportional to the pyrophosphate present. This assay is simpler and more robust than traditional enzyme-based methods and is ideal for screening enzyme activity or enzyme inhibitors.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service