1. There is no specific recommended amount of protein per well for testing. It is best to optimize this by testing a range of dilutions to determine the most effective concentration for the specific sample being used.
2. The assay buffer does not contain any inhibitors.
3. Mild sonication can be used instead of a dounce homogenizer, with low amplitude and short pulses, while keeping the samples on ice throughout the sonication process.
MAK356
Indoleamine 2,3-Dioxygenase 1 (IDO1) Activity Assay Kit
sufficient for 100 fluorometric tests
Synonym(s):
IDO1 Activity Assay Kit
About This Item
Recommended Products
usage
sufficient for 100 fluorometric tests
detection method
fluorometric
relevant disease(s)
immunological diseases; cancer
General description
Features and Benefits
Suitability
Principle
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
188.6 °F
Flash Point(C)
87 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
1. What is the recommended concentration of protein to use per well?
2. Do the IDO kit buffers contain inhibitors for endogenous enzymes that may metabolize N-formylkynurinine?
3. Will anything in the kit be affected by using sonication instead of a dounce homogenizer?1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service