Skip to Content
Merck
All Photos(1)

Key Documents

SAE0096

Sigma-Aldrich

IDO1, human

recombinant, expressed in E. coli, lyophilized powder, His tagged

Synonym(s):

Indoleamine 2,3-dioxygenase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26
Pricing and availability is not currently available.

recombinant

expressed in E. coli

form

lyophilized powder

specific activity

800 units/μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

General description

IDO1 is a heme-enzyme that is induced by INF-γ and catalyzes the conversion of tryptophan to kynurenine. This leads to a depletion of tryptophan that results in T cells and dendritic cells inactivation. IDO activity has been identified as an important immune effector pathway in tumor cells to escape a potentially effective immune response. Inhibition of IDO1 has been found to delay cancer growth and enhance dendritic vaccines. Therapeutic inhibition of IDO1 is of interest because of its proposed role in the pathogenesis of several diseases, including cancer, hypotension, neurodegenerative disorders, and its potential to aid cancer immunotherapy and organ transplant.
This recombinant human IDO1 has a 6x histidine tag at its N-terminus. This product is lyophilized from a solution containing 10 mM Trizma buffer (pH 7.0), with 500 mM NaCl and a carbohydrate carrier.

Unit Definition

One unit of enzyme hydrolyzes 1.0 pmole of L-Tryptophan to N-formyl-Lkynurenine per minute at pH-6.5 at 37°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immune-modulating enzyme indoleamine 2,3-dioxygenase is effectively inhibited by targeting its apo-form
Nelp, M.T. et al.
Proceedings of the National Academy of Sciences of the USA, 115(13), 3249-3254 (2018)
Indoleamine 2,3-dioxygenase: is it an immune suppressor?
Sollman, H. et al.
Cancer Journal, 16(4), 354-359 (2010)
Ji-Hee Shin et al.
European journal of nutrition, 59(8), 3591-3601 (2020-02-15)
Growing evidence shows that nutrient metabolism affects inflammatory bowel diseases (IBD) development. Previously, we showed that deficiency of indoleamine 2,3-dioxygenase 1 (Ido1), a tryptophan-catabolizing enzyme, reduced the severity of dextran sulfate sodium (DSS)-induced colitis in mice. However, the roles played
Human bone marrow stromal cells inhibit allogeneic T-cell responses by indoleamine 2,3-dioxygenase-mediated tryptophan degradation.
Meisel, R. et al.
Blood, 103(12), 4619-4621 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service