Skip to Content
Merck
All Photos(1)

Documents

M7687

Greiner 96 well 1 x 8 strip plates

medium binding, flat bottom clear polystyrene wells (8 well strips)

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15

material

clear polystyrene
flat bottom clear polystyrene wells (8 well strips)

sterility

non-sterile

feature

lid: no

packaging

case of 100

manufacturer/tradename

Greiner 762070

maximum volume

388 μL

size

96 wells

working volume

350 μL

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

MICROLON® 200, 96W 12xF8 Strip Microplate, working volume 20-350 μL; total well volume 388 μL.

Other Notes

96 well flat (F) solid bottom microplate in white frame, F8 strips mounted in frame, medium-binding, no lid, alphanumeric well coding, length: 127.76 mm; width: 85.48 mm.

Suitability

UsefμL in immunological assays. MICROLON 200 is a hydrophobic medium binding surface that will bind up to 200 ng/cm2 of non-polar proteins and peptides. Free of detectable DNase/RNase, human DNA and pyrogens.

Legal Information

MICROLON is a registered trademark of Greiner Bio-One GmbH

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service