Key Documents
BR782301
BRAND® strip plates
size 96 wells, 12xF8, MB, dividable: yes
Synonym(s):
BRANDplates®, Micro Tubes with Screw caps, Microcentrifuge Tubes, Microplates, Plates, Screw caps, Strip Plates
Select a Size
100 EA
$1,350.00
$1,350.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
100 EA
$1,350.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
material:
polystyrene
transparent wells
size:
96 wells
sterility:
non-sterile
binding type:
medium binding surface
feature:
12xF8, MB
pureGrade
$1,350.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polystyrene
transparent wells
sterility
non-sterile
feature
12xF8, MB
pureGrade
packaging
pack of 100 ea
pkg of (4 bags of 25)
manufacturer/tradename
BRAND 782301
size
96 wells
binding type
medium binding surface
General description
  • BRAND® strip plates are polystyrene, transparent with grid, strips of 8 wells, divisible and flat bottom.
  • Optimal surfaces enable optimal results.
  • Reliable quality and tested by independent laboratories.
  • Ideal visual properties and excellent flexibility.
  • In accordance with ANSI/SLAS production standards.
Features and Benefits
  • Optimal surfaces for optimal results
  • Reliable quality from the cleanroom
  • Tested by independent laboratories
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)