Skip to Content
Merck
All Photos(1)

Key Documents

M5131

Sigma-Aldrich

MTH-DL-Methionine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2OS2
CAS Number:
Molecular Weight:
204.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Pricing and availability is not currently available.

storage temp.

2-8°C

SMILES string

CSCCC1NC(=S)N(C)C1=O

InChI

1S/C7H12N2OS2/c1-9-6(10)5(3-4-12-2)8-7(9)11/h5H,3-4H2,1-2H3,(H,8,11)

InChI key

GQSMFARWFJRERX-UHFFFAOYSA-N

Biochem/physiol Actions

MTH-DL-Methionine is a methylthiohydantoin amino acid derivative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid-Chromatographic separation with Electrochemical Detection of the Phenylthiohydantoin and Methylthiohydantoin Derivatives of Amino-Acids
WELCH LE; JOHNSON DC
Journal of Chromatography A, 180, 111-118 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service