Skip to Content
Merck
All Photos(1)

Key Documents

M2922

Sigma-Aldrich

3-Methyl-6-[3-(trifluoromethyl)phenyl]-1,2,4-triazolo[4,3-b]pyridazine

Synonym(s):

CL 218,872

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9F3N4
CAS Number:
Molecular Weight:
278.23
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

Cc1nnc2ccc(nn12)-c3cccc(c3)C(F)(F)F

Biochem/physiol Actions

Selective agonist at the BZI benzodiazepine receptor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Karolak-Wojciechowska et al.
Bioorganic & medicinal chemistry, 2(8), 773-779 (1994-08-01)
Electronic parameters (molecular electrostatic potential MEP, charge distribution on the nitrogen atoms, dipole moment mu and ionization potential IP) were calculated by semiempirical quantum chemistry methods for 2 sets (X = H and m-CF3, the syn- and anti-rotamers of the
R K McNamara et al.
Epilepsy research, 16(1), 19-26 (1993-09-01)
To clarify the role of benzodiazepine receptors in kindling, the present experiment assessed the effects of CL 218,872 (1, 5, 10, and 20 mg/kg), a triazolopyridazine with a selective affinity for the putative benzodiazepine BZ1 receptor subtype, on the development
H K Im et al.
Molecular pharmacology, 44(4), 866-870 (1993-10-01)
Previous studies with cloned gamma-aminobutyric acid type A receptors expressed in human embryonic kidney cells have indicated that the alpha 1 beta 2 gamma 2 and alpha 1 gamma 2 (but not alpha 1 beta 2) subtypes have benzodiazepine sites.
O Giorgi et al.
European journal of pharmacology, 290(1), 37-47 (1995-06-23)
The present study was designed to compare the allosteric coupling between the Cl- channel of the GABAA receptor and the different benzodiazepine recognition site subtypes (BZ sites) in the cerebral cortex of newborn (5-day-old) and adult rats (90-day-old). To this
R L Hyson et al.
Journal of molecular neuroscience : MN, 8(3), 193-205 (1997-06-01)
Neurons in nucleus magnocellularis (NM) and nucleus laminaris (NL) of the chick brainstem auditory system show an unusual physiological response to GABA. Examination of these nuclei using in situ hybridization for GABAA receptor subunits showed a differential expression of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service