Skip to Content
Merck
All Photos(1)

Documents

M1949

Sigma-Aldrich

S-methyl-5′-thioadenosine phosphorylase human

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

Assay

≥80% (SDS-PAGE)

form

solution

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−70°C

Application

S-methyl-5′-thioadenosine phosphorylase human (MTAP) is an enzyme used in cancer research that is deficient in many types of cancer. Decreased MTAP expression may be used as a potential indicator of disease progression of gastrointestinal stromal tumors . MTAP may be a used to develop potential therapeutic strategies for hepatocellular carcinoma (HCC) since MTAP inactivation has been linked to HCC development and invasiveness .

Biochem/physiol Actions

MTAP expression is crucial for the catabolism of methylthioadenosine, which is a by-product of polyamine biosynthesis in the methionine salvage pathway. Protein expression is decreased by homozygous deletion and promoter hypermethylation .

Physical properties

N-terminal GST-tagged 57 kDa protein containing amino acids 2-end.

Physical form

Supplied as a solution in 25 mM Tris-HCl, pH 8.0,100 mM NaCl, 0.05% Tween®-20, 10% glycerol,and 3 mM DTT.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tony W H Li et al.
Carcinogenesis, 33(2), 427-435 (2011-12-14)
Chronic inflammation is an underlying risk factor for colon cancer. Tumor necrosis factor alpha (TNF-α) plays a critical role in the development of inflammation-induced colon cancer in a mouse model. S-adenosylmethionine (SAMe) and its metabolite methylthioadenosine (MTA) can inhibit lipopolysaccharide-induced
Zarah Glad Zimling et al.
Histopathology, 60(6B), E96-105 (2012-03-08)
Malignant pleural mesothelioma (MPM) often causes diagnostic difficulties for pathologists. We assessed whether loss of methylthioadenosine phosphorylase (MTAP), a key enzyme in the intracellular recycling of adenosine triphosphate (ATP) often deleted in MPM, could be detected with immunohistochemistry (IHC) and
Promoter-hypermethylation is causing functional relevant downregulation of methylthioadenosine phosphorylase (MTAP) expression in hepatocellular carcinoma
Claus Hellerbrand, Marcus Muhlbauer, et al.
Molecular Pharmacology, 52, 64-72 (2006)
Shasha Li et al.
Zhongguo fei ai za zhi = Chinese journal of lung cancer, 14(2), 151-155 (2011-02-24)
The abnormal expression of MTAP, a tumor suppressor gene, is found in a variety of tumor tissues. The aim of this study is to detect the expression of MTAP mRNA protein and the clinical significance for the therapy of non-small
Hsuan-Ying Huang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 15(22), 6963-6972 (2009-11-06)
Chromosome 9 is frequently deleted in high-risk gastrointestinal stromal tumors (GISTs), whereas its specific tumor suppressor genes (TSGs) are less understood. We did an integrative study of MTAP gene at 9p21 to analyze its implication in GISTs. To search TSGs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service