Skip to Content
Merck
All Photos(1)

Key Documents

FNCAS9PROT

Sigma-Aldrich

FnCas9 Protein

from Francisella novicida, recombinant, expressed in E. coli, 1X NLS

Synonym(s):

Francisella Novicida Cas9

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$138.00
250 μG
$502.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$138.00
250 μG
$502.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.51

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

packaging

pkg of 1 kit (4 components)

application(s)

CRISPR

shipped in

wet ice

storage temp.

−20°C

General description

The purified FnCas9 protein (~190 kDa) is derived from the type II CRISPR/Cas system from the bacterium Francisella novicida and has been engineered to function using three molecular components: the FnCas9 protein, a crRNA of user-defined sequence, and a tracrRNA of constant sequence. The FnCas9 endonuclease can be programmed by changing the crRNA sequence to create a site-specific double-strand break (DSB) in plasmids or purified genomic DNA.

Application

Functional Genomics/Target Validation/Genome Editing/Molecular Biology/Molecular Cloning/Synthetic Biology

Features and Benefits

  • Suitable for plasmid or large genomic DNA fragment cloning
  • Results in 4-bp 5′ overhangs.
  • Cuts purified plasmid DNA more efficiently than SpCas9

Packaging

pkg of 50 μg (≥ 300 pmol)
pkg of 250 μg (≥ 1500 pmol)

Components

Each kit consists of:
  • one vial of FnCas9 recombinant protein
  • one vial containing 1 mL of 1× Dilution buffer
  • one vial containing 1 mL of Nuclease free water with glycerol
  • one vial containing 1 mL of 10× Digestion Buffer

Principle

Conventional restriction enzymes such as EcoRI and HindIII can only cut specific short (6 bp) DNA sequences and are therefore limited to cloning small DNA fragments (typically ~4 kb). CRISPR-based endonculeases can cut user defined sequences of a longer length (~20 bp) making them more flexible and applicable to larger DNA fragments such as multigene bacterial operons or mammalian genomic DNA with native promoter, exon, and intron components.

Reconstitution

Lyophilized FnCas9 protein should be resuspended in the Reconstitution solution provided to desired concentration. Gently tap tube to completely dissolve lyophilized powder, incubate for 10 minutes on ice, and spin tube to bring material to bottom of tube.

Other Notes

Click here to order gRNA
Enter the 20bp genomic DNA target sequence (5′ to 3′) plus the adjacent 3′ PAM site. Under special instructions, indicate "FnCas9 crRNA"
Click here to order FnCas9 tracrRNA

Check out our other MISSION® Cas9 Proteins at SigmaAldrich.com/CRISPRproteins

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Only

Product No.
Description

  • FnCas9-NLS from Francisella novicida, expressed in Escherichia coli

  • Dilution buffer for Cas9 proteins

  • Reconstitution solution for Cas9 proteins

  • Digestion buffer 10× for FnCas9

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Combine guaranteed sgRNAs with our comprehensive range of CRISPR products and tools, including Cas9 and delivery reagents, for efficient genome modification with higher specificity.

Related Content

Combine guaranteed sgRNAs with our comprehensive range of CRISPR products and tools, including Cas9 and delivery reagents, for efficient genome modification with higher specificity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service