Skip to Content
Merck
All Photos(1)

Key Documents

M110

Sigma-Aldrich

α-Methylserotonin maleate salt

≥98% (HPLC)

Synonym(s):

α-Methyl-5-hydroxytryptamine maleate salt, (±)-3-(2-Aminopropyl)indol-5-ol maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O · C4H4O4
CAS Number:
Molecular Weight:
306.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: >3 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.CC(N)Cc1c[nH]c2ccc(O)cc12

InChI

1S/C11H14N2O.C4H4O4/c1-7(12)4-8-6-13-11-3-2-9(14)5-10(8)11;5-3(6)1-2-4(7)8/h2-3,5-7,13-14H,4,12H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

YQNHFSXRABPJLP-BTJKTKAUSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

5-HT2 serotonin receptor agonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ralf Buettner et al.
Molecular therapy oncolytics, 14, 149-158 (2019-06-19)
The immunosuppressive agent leflunomide has been used in the treatment of over 300,000 patients with rheumatoid arthritis. Its active metabolite, teriflunomide (Ter), directly inhibits dihydroorotate dehydrogenase (DHODH), an enzyme involved in nucleoside synthesis. We report that Ter not only shows
P D Feldman
Journal of the autonomic nervous system, 56(1-2), 119-124 (1995-12-05)
Spontaneous neuronal activity in the solitary tract nucleus was recorded extracellularly in a brain slice preparation during bath-application of 5-HT1 and 5-HT2 receptor-selective agonists and antagonists. The 5-HT1A/5-HT1B agonist 5-carboxamidotryptamine depressed activity in 20 of 25 neurons studied. The remaining
A M Ismaiel et al.
Journal of medicinal chemistry, 33(2), 755-758 (1990-02-01)
alpha-Methyl-5-hydroxytryptamine (alpha-Me-5-HT; 2) and 2-methyl-5-hydroxytryptamine (2-Me-5-HT; 3) are considered to be 5-HT2-selective and 5-HT3-selective agents, respectively. These agents were synthesized and examined at serotonin (5-HT) binding sites because there is relatively little documentation as to their selectivity and because they
G P Leung et al.
The Journal of physiology, 519 Pt 3, 657-667 (1999-08-24)
1. The short-circuit current (Isc) technique was used to study the role of 5-hydroxytryptamine (5-HT) in the regulation of anion secretion in cultured rat cauda epididymal epithelia. 2. 5-HT, the 5-HT1B-selective agonist 5-nonyloxytryptamine (5-NOT) and the 5-HT2B-selective agonist alpha-methyl-5-hydroxytryptamine (alpha-methyl-5-HT)
Dmitrii Zabelskii et al.
Nature communications, 11(1), 5707-5707 (2020-11-13)
Phytoplankton is the base of the marine food chain as well as oxygen and carbon cycles and thus plays a global role in climate and ecology. Nucleocytoplasmic Large DNA Viruses that infect phytoplankton organisms and regulate the phytoplankton dynamics encompass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service