Skip to Content
Merck
All Photos(1)

Key Documents

B4063

Sigma-Aldrich

BIMU8 hydrate

≥98% (HPLC)

Synonym(s):

2,3-Dihydro-N-[(3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]-3-(1-methylethyl)-2-oxo-1H-benzimidazole-1-carboxamide Hydrochloride (1:1) hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$302.00

$302.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$302.00

About This Item

Empirical Formula (Hill Notation):
C19H26N4O2·HCl · xH2O
CAS Number:
Molecular Weight:
378.90 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$302.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

color

off-white to light tan

solubility

H2O: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

Cl.N1([C@@H]2C[C@H](C[C@H]1CC2)NC(=O)N3c4c(cccc4)N(C3=O)C(C)C)C.O

InChI

1S/C19H26N4O2.ClH.H2O/c1-12(2)22-16-6-4-5-7-17(16)23(19(22)25)18(24)20-13-10-14-8-9-15(11-13)21(14)3;;/h4-7,12-15H,8-11H2,1-3H3,(H,20,24);1H;1H2/t13-,14+,15-;;

InChI key

HZJJVFOOACXPTH-XZAJHMFNSA-N

Related Categories

Biochem/physiol Actions

BIMU8 hydrate is a potent 5-HT4 serotonin receptor agonist.
BIMU8 hydrate is a potent 5-HT4 serotonin receptor agonist. Serotonin (5-HT) is a major neurotransmitter that acts through a family of GPCRs and one ion channel. 5-HT4 receptor is GPCR expressed in many tissues, including brain, and modulates dopamine secretion, learning, and memory. BIMU8 is a full agonist at 5-HT4, but it binds differently than the endogenous ligand, 5-HT, shown through site-directed mutagenesis studies. It depolarizes neurons and was used to localize 5-HT4 to somatic but not dendritic regions of CA1 pyramidal neurons.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Contesse et al.
European journal of pharmacology, 265(1-2), 27-33 (1994-11-14)
We have previously shown that serotonin (5-hydroxytryptamine, 5-HT) stimulate corticosterone and aldosterone secretion from perifused frog adrenal gland in vitro through activation of 5-HT4 receptors. In the present study, we have used this model to investigate the effect of newly
A Dumuis et al.
Naunyn-Schmiedeberg's archives of pharmacology, 345(3), 264-269 (1992-03-01)
Three chemical classes of serotonin 5-HT4 receptor agonists have been identified so far: 5-substituted indoles (e.g. 5-HT), benzamides (e.g. renzapride) and benzimidazolones (e.g. BIMU 8). In a search for 5-HT4 receptor antagonists, we have discovered that the benzimidazolone derivative DAU
A Dumuis et al.
Naunyn-Schmiedeberg's archives of pharmacology, 343(3), 245-251 (1991-03-01)
Recent experimental evidence indicates that central 5-HT4 receptors which are positively coupled to adenylate cyclase, are stimulated by a family of 2-methoxy-4-amino-5-chloro substituted benzamide derivatives. These compounds are also potent stimulants of the gastro-intestinal motility. In this study the ability
H Kilbinger et al.
Naunyn-Schmiedeberg's archives of pharmacology, 351(3), 229-236 (1995-03-01)
The effects of the 5-HT4 receptor agonists BIMU 8, BIMU 1, renzapride and of the 5-HT1p receptor agonist 5-hydroxyindalpine on basal and electrically evoked outflow of tritium were studied in guinea-pig longitudinal muscle myenteric plexus preparations preincubated with [3H]choline. Muscle
J K Weatherspoon et al.
European journal of pharmacology, 326(2-3), 133-138 (1997-05-20)
The binding profile of the sigma2 receptor ligand endo-N-(8-methyl-8-azabicyclo[3.2.1.]oct-3-yl)-2,3-dihydro-(1-methyl)eth yl-2-oxo-1H-benzimidazole-1-carboxamidehydrochloride (BIMU-8) had previously been determined, but its agonist/antagonist status at sigma2 receptors had not been identified. We therefore investigated the effects of BIMU-8 for its ability to regulate the stimulated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service