Skip to Content
Merck
All Photos(1)

Documents

M0325

Sigma-Aldrich

Minimum Essential Medium Eagle

With Earle′s salts, non-essential amino acids, ʟ-glutamine and sodium bicarbonate, sterile liquid, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

glucose: 1.0 g/L (Dextro)
L-glutamine: 0.292 g/L
phenol red: 0.011 g/L
Earle’s salts (5% CO2): yes
NaHCO3: 2.2 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Minimum Essential Medium Eagle (MEM) is a synthetic cell culture media, developed by Harry Eagle. It is used for the cultivation of a wide variety of cells grown in monolayers. MEM includes higher concentrations of amino acids so the medium more closely approximates the protein composition of cultured mammalian cells. May be preferred for the growth of adherent cells in the presence of reduced serum or as a base for the development of a serum-free medium.

Application

Minimum Essential Medium Eagle has been used as a culture medium for:
  • thyroid cancer cell lines
  • macrophage-like DH82 cells
  • vero CCL-81 cells

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sharanjeet Atwal et al.
mBio, 12(4), e0134221-e0134221 (2021-07-28)
Peptidoglycan (PG) is a highly cross-linked peptide-glycan mesh that confers structural rigidity and shape to most bacterial cells. Polymerization of new PG is usually achieved by the concerted activity of two membrane-bound machineries, class-A penicillin binding proteins (aPBPs) and class-B
Sharanjeet Atwal et al.
Journal of microbiological methods, 169, 105812-105812 (2019-12-22)
Despite their clinical and biological importance, the cell biology of obligate intracellular bacteria is less well understood than that of many free-living model organisms. One reason for this is that they are mostly genetically intractable. As a consequence, it is
Fang Feng et al.
Cancer research, 78(21), 6121-6133 (2018-09-16)
The sodium iodide symporter (NIS) is a classical iodide pump typically localized within the cell plasma membrane in thyroid cells, where NIS expression is believed to ensure success of mainstay radioiodide therapy in thyroid cancers. Although radioiodide uptake is generally
Jose Manuel Condor Capcha et al.
Frontiers in cardiovascular medicine, 7, 618651-618651 (2021-02-02)
The COVID-19 pandemic caused by the SARS-CoV-2 coronavirus requires reliable assays for studying viral entry mechanisms which remains poorly understood. This knowledge is important for the development of therapeutic approaches to control SARS-CoV-2 infection by permitting the screening for neutralizing
Ana Campos Codo et al.
Cell metabolism, 32(3), 437-446 (2020-07-23)
COVID-19 can result in severe lung injury. It remained to be determined why diabetic individuals with uncontrolled glucose levels are more prone to develop the severe form of COVID-19. The molecular mechanism underlying SARS-CoV-2 infection and what determines the onset

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service